Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45101122-45101367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUOX2
Entrez Gene ID
50506
Gene Name
dual oxidase 2
Gene Aliases
LNOX2, NOXEF2, P138-TOX, TDH6, THOX2
UniGene
Hs.71377
Gene Chromosome Location
Chr.15: 45092653-45114161 on Build GRCh38
Transcripts
NM_014080.4 XM_005254421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1684402 c.2893_2896delTCGT p.F966fs*29 Chr.15: 45101230-45101233 on Build GRCh38
3501628 c.2894C>T p.S965L Chr.15: 45101232-45101232 on Build GRCh38
4807262 c.2914G>T p.G972W Chr.15: 45101212-45101212 on Build GRCh38
5643695 c.2905C>G p.R969G Chr.15: 45101221-45101221 on Build GRCh38
3401764 c.2906G>A p.R969Q Chr.15: 45101220-45101220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019842944 Chr.15: 45101327-45101327 on Build GRCh38
rs138726481 Chr.15: 45101332-45101332 on Build GRCh38
rs375112603 Chr.15: 45101221-45101221 on Build GRCh38
rs991144438 Chr.15: 45101326-45101326 on Build GRCh38
rs772117868 Chr.15: 45101307-45101307 on Build GRCh38
rs747046672 Chr.15: 45101311-45101311 on Build GRCh38
rs373159143 Chr.15: 45101189-45101189 on Build GRCh38
rs774685584 Chr.15: 45101211-45101211 on Build GRCh38
rs775652056 Chr.15: 45101310-45101310 on Build GRCh38
rs766056568 Chr.15: 45101238-45101238 on Build GRCh38
rs763248481 Chr.15: 45101188-45101189 on Build GRCh38
rs111706445 Chr.15: 45101163-45101163 on Build GRCh38
rs146664125 Chr.15: 45101220-45101220 on Build GRCh38
rs200985557 Chr.15: 45101243-45101243 on Build GRCh38
rs145038941 Chr.15: 45101261-45101261 on Build GRCh38
rs199783724 Chr.15: 45101239-45101239 on Build GRCh38
rs761813221 Chr.15: 45101224-45101224 on Build GRCh38
rs759739622 Chr.15: 45101219-45101219 on Build GRCh38
rs752093078 Chr.15: 45101262-45101262 on Build GRCh38
rs369899254 Chr.15: 45101179-45101179 on Build GRCh38
rs144153950 Chr.15: 45101232-45101232 on Build GRCh38
rs369286028 Chr.15: 45101294-45101294 on Build GRCh38
rs778216481 Chr.15: 45101205-45101205 on Build GRCh38
rs146077650 Chr.15: 45101170-45101170 on Build GRCh38
rs757674714 Chr.15: 45101287-45101287 on Build GRCh38
rs771346442 Chr.15: 45101209-45101209 on Build GRCh38
rs765025859 Chr.15: 45101226-45101226 on Build GRCh38
rs770135491 Chr.15: 45101193-45101193 on Build GRCh38
rs974087064 Chr.15: 45101297-45101297 on Build GRCh38
rs749710666 Chr.15: 45101269-45101269 on Build GRCh38
rs530719719 Chr.15: 45101228-45101231 on Build GRCh38
rs1008911749 Chr.15: 45101304-45101304 on Build GRCh38
rs758060177 Chr.15: 45101162-45101162 on Build GRCh38
rs781596874 Chr.15: 45101183-45101183 on Build GRCh38
rs892098421 Chr.15: 45101273-45101273 on Build GRCh38
rs754451450 Chr.15: 45101173-45101173 on Build GRCh38
rs780176601 Chr.15: 45101345-45101345 on Build GRCh38
rs1053440250 Chr.15: 45101185-45101185 on Build GRCh38
rs549117333 Chr.15: 45101321-45101321 on Build GRCh38
rs900037641 Chr.15: 45101336-45101336 on Build GRCh38
rs778142275 Chr.15: 45101268-45101268 on Build GRCh38
rs775863038 Chr.15: 45101218-45101219 on Build GRCh38
rs982742385 Chr.15: 45101143-45101143 on Build GRCh38
rs756705365 Chr.15: 45101266-45101266 on Build GRCh38
rs140446258 Chr.15: 45101231-45101231 on Build GRCh38
rs749661602 Chr.15: 45101207-45101207 on Build GRCh38
rs199968378 Chr.15: 45101290-45101290 on Build GRCh38

View Full Product Details