Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55519703-55519966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP36
Entrez Gene ID
112942
Gene Name
cilia and flagella associated protein 36
Gene Aliases
BARTL1, CCDC104
UniGene
Hs.264208
Gene Chromosome Location
Chr.2: 55519595-55545080 on Build GRCh38
Transcripts
NM_001282761.1 NM_080667.6 XM_017003254.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4094907 c.58C>A p.L20M Chr.2: 55519859-55519859 on Build GRCh38
5567909 c.76T>C p.S26P Chr.2: 55519877-55519877 on Build GRCh38
4392541 c.32G>T p.W11L Chr.2: 55519833-55519833 on Build GRCh38
443046 c.41A>G p.E14G Chr.2: 55519842-55519842 on Build GRCh38
4094906 c.58C>A p.L20M Chr.2: 55519859-55519859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779945399 Chr.2: 55519811-55519813 on Build GRCh38
rs762063763 Chr.2: 55519838-55519838 on Build GRCh38
rs372668203 Chr.2: 55519851-55519851 on Build GRCh38
rs765019088 Chr.2: 55519760-55519760 on Build GRCh38
rs747658569 Chr.2: 55519815-55519815 on Build GRCh38
rs762692170 Chr.2: 55519834-55519834 on Build GRCh38
rs754366142 Chr.2: 55519844-55519844 on Build GRCh38
rs766469429 Chr.2: 55519944-55519944 on Build GRCh38
rs191718325 Chr.2: 55519865-55519865 on Build GRCh38
rs551462553 Chr.2: 55519825-55519825 on Build GRCh38
rs756242029 Chr.2: 55519842-55519842 on Build GRCh38
rs749455352 Chr.2: 55519753-55519754 on Build GRCh38
rs372788631 Chr.2: 55519774-55519774 on Build GRCh38
rs540461610 Chr.2: 55519742-55519742 on Build GRCh38
rs574366687 Chr.2: 55519779-55519779 on Build GRCh38
rs752393299 Chr.2: 55519766-55519766 on Build GRCh38
rs780701711 Chr.2: 55519866-55519866 on Build GRCh38
rs201464473 Chr.2: 55519789-55519789 on Build GRCh38
rs1029964500 Chr.2: 55519725-55519725 on Build GRCh38
rs578131577 Chr.2: 55519724-55519724 on Build GRCh38
rs763941107 Chr.2: 55519835-55519835 on Build GRCh38
rs777356694 Chr.2: 55519805-55519805 on Build GRCh38
rs567259044 Chr.2: 55519931-55519931 on Build GRCh38
rs762151425 Chr.2: 55519920-55519920 on Build GRCh38
rs750485058 Chr.2: 55519841-55519841 on Build GRCh38
rs200835926 Chr.2: 55519797-55519797 on Build GRCh38
rs746668375 Chr.2: 55519863-55519863 on Build GRCh38
rs781366585 Chr.2: 55519783-55519783 on Build GRCh38
rs775474019 Chr.2: 55519826-55519826 on Build GRCh38
rs776607466 Chr.2: 55519755-55519755 on Build GRCh38
rs897986815 Chr.2: 55519868-55519868 on Build GRCh38
rs974295889 Chr.2: 55519757-55519757 on Build GRCh38
rs560814667 Chr.2: 55519750-55519750 on Build GRCh38
rs918761843 Chr.2: 55519761-55519761 on Build GRCh38
rs755357181 Chr.2: 55519853-55519853 on Build GRCh38
rs985701240 Chr.2: 55519744-55519744 on Build GRCh38
rs376599393 Chr.2: 55519804-55519804 on Build GRCh38
rs749231499 Chr.2: 55519889-55519889 on Build GRCh38
rs547165610 Chr.2: 55519878-55519878 on Build GRCh38
rs560315196 Chr.2: 55519870-55519870 on Build GRCh38
rs773538975 Chr.2: 55519936-55519936 on Build GRCh38
rs376313385 Chr.2: 55519788-55519788 on Build GRCh38
rs779278243 Chr.2: 55519854-55519854 on Build GRCh38
rs745834830 Chr.2: 55519821-55519821 on Build GRCh38
rs79501524 Chr.2: 55519941-55519941 on Build GRCh38
rs776050268 Chr.2: 55519820-55519820 on Build GRCh38
rs749665952 Chr.2: 55519798-55519798 on Build GRCh38
rs774160357 Chr.2: 55519916-55519916 on Build GRCh38
rs746659408 Chr.2: 55519814-55519814 on Build GRCh38
rs750973554 Chr.2: 55519792-55519792 on Build GRCh38
rs758141098 Chr.2: 55519786-55519787 on Build GRCh38
rs903020337 Chr.2: 55519942-55519942 on Build GRCh38
rs764269561 Chr.2: 55519776-55519776 on Build GRCh38
rs774453282 Chr.2: 55519837-55519837 on Build GRCh38
rs768344640 Chr.2: 55519900-55519900 on Build GRCh38
rs369074330 Chr.2: 55519810-55519810 on Build GRCh38
rs767740011 Chr.2: 55519839-55519839 on Build GRCh38
rs751770746 Chr.2: 55519777-55519777 on Build GRCh38
rs759410915 Chr.2: 55519759-55519759 on Build GRCh38
rs34891804 Chr.2: 55519819-55519819 on Build GRCh38
rs766837635 Chr.2: 55519843-55519843 on Build GRCh38
rs199958381 Chr.2: 55519871-55519871 on Build GRCh38

View Full Product Details