Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 17774858-17775095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF13A
Entrez Gene ID
63971
Gene Name
kinesin family member 13A
Gene Aliases
RBKIN, bA500C11.2
UniGene
Hs.94499
Gene Chromosome Location
Chr.6: 17759183-17987623 on Build GRCh38
Transcripts
NM_001105566.2 NM_001105567.2 NM_001105568.2 NM_022113.5 XM_017011188.1 XM_017011189.1 XM_017011190.1 XM_017011191.1 XM_017011192.1 XM_017011193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4766543 c.4192C>T p.L1398F Chr.6: 17775041-17775041 on Build GRCh38
7373775 c.4148C>T p.P1383L Chr.6: 17775046-17775046 on Build GRCh38
4766542 c.4153C>T p.L1385F Chr.6: 17775041-17775041 on Build GRCh38
7373774 c.4187C>T p.P1396L Chr.6: 17775046-17775046 on Build GRCh38
5779924 c.4175C>G p.S1392C Chr.6: 17775058-17775058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370143886 Chr.6: 17774960-17774960 on Build GRCh38
rs748032031 Chr.6: 17774997-17774997 on Build GRCh38
rs770047708 Chr.6: 17775022-17775022 on Build GRCh38
rs376404952 Chr.6: 17775008-17775008 on Build GRCh38
rs569146131 Chr.6: 17775032-17775032 on Build GRCh38
rs200785773 Chr.6: 17774990-17774990 on Build GRCh38
rs755135919 Chr.6: 17774967-17774967 on Build GRCh38
rs900971113 Chr.6: 17775050-17775050 on Build GRCh38
rs571068042 Chr.6: 17774947-17774947 on Build GRCh38
rs763389412 Chr.6: 17775034-17775034 on Build GRCh38
rs759983587 Chr.6: 17775018-17775018 on Build GRCh38
rs200833262 Chr.6: 17775046-17775046 on Build GRCh38
rs886653773 Chr.6: 17775024-17775024 on Build GRCh38
rs769523306 Chr.6: 17775007-17775007 on Build GRCh38
rs775643529 Chr.6: 17774977-17774978 on Build GRCh38
rs200901764 Chr.6: 17775049-17775049 on Build GRCh38
rs754710081 Chr.6: 17774983-17774983 on Build GRCh38
rs779859137 Chr.6: 17774979-17774979 on Build GRCh38
rs917252776 Chr.6: 17774935-17774935 on Build GRCh38
rs150868581 Chr.6: 17774981-17774981 on Build GRCh38
rs771227807 Chr.6: 17775006-17775006 on Build GRCh38
rs557099674 Chr.6: 17775004-17775004 on Build GRCh38
rs988787555 Chr.6: 17774910-17774910 on Build GRCh38
rs756978744 Chr.6: 17775055-17775055 on Build GRCh38
rs777933544 Chr.6: 17775002-17775002 on Build GRCh38
rs970851431 Chr.6: 17774932-17774933 on Build GRCh38
rs772335874 Chr.6: 17775000-17775000 on Build GRCh38
rs930793282 Chr.6: 17774987-17774987 on Build GRCh38
rs753403109 Chr.6: 17775062-17775062 on Build GRCh38
rs1025540967 Chr.6: 17774894-17774895 on Build GRCh38
rs371587242 Chr.6: 17775045-17775045 on Build GRCh38
rs755701451 Chr.6: 17774978-17774978 on Build GRCh38
rs767024112 Chr.6: 17775042-17775042 on Build GRCh38
rs945886859 Chr.6: 17775060-17775060 on Build GRCh38

View Full Product Details