Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 68539232-68539505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM20A
Entrez Gene ID
54757
Gene Name
family with sequence similarity 20 member A
Gene Aliases
AI1G, AIGFS, FP2747
UniGene
Hs.268874
Gene Chromosome Location
Chr.17: 68534561-68600954 on Build GRCh38
Transcripts
NM_001243746.1 NM_017565.3 NR_027751.1 XM_006721959.2 NM_001276290.1

Gene Symbol
PRKAR1A
Entrez Gene ID
5573
Gene Name
protein kinase cAMP-dependent type I regulatory subunit alpha
Gene Aliases
ACRDYS1, ADOHR, CAR, CNC, CNC1, PKR1, PPNAD1, PRKAR1, TSE1
UniGene
Hs.280342
Gene Chromosome Location
Chr.17: 68413623-68551316 on Build GRCh38
Transcripts
NM_001243746.1 NM_017565.3 NR_027751.1 XM_006721959.2 NM_001276290.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1303249 c.1324G>A p.E442K Chr.17: 68539374-68539374 on Build GRCh38
7135920 c.1314C>A p.H438Q Chr.17: 68539384-68539384 on Build GRCh38
983431 c.1340C>T p.S447L Chr.17: 68539358-68539358 on Build GRCh38
6659397 c.1305C>T p.F435F Chr.17: 68539393-68539393 on Build GRCh38
7232464 c.1361+8T>C p.? Chr.17: 68539329-68539329 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs533347978 Chr.17: 68539310-68539310 on Build GRCh38
rs773465041 Chr.17: 68539293-68539293 on Build GRCh38
rs757155258 Chr.17: 68539330-68539330 on Build GRCh38
rs770194120 Chr.17: 68539388-68539388 on Build GRCh38
rs763888844 Chr.17: 68539314-68539314 on Build GRCh38
rs373199897 Chr.17: 68539286-68539286 on Build GRCh38
rs890901752 Chr.17: 68539308-68539308 on Build GRCh38
rs200391198 Chr.17: 68539371-68539371 on Build GRCh38
rs759494182 Chr.17: 68539432-68539432 on Build GRCh38
rs745467675 Chr.17: 68539473-68539473 on Build GRCh38
rs867364658 Chr.17: 68539259-68539259 on Build GRCh38
rs1044385002 Chr.17: 68539466-68539466 on Build GRCh38
rs779624390 Chr.17: 68539358-68539358 on Build GRCh38
rs766452056 Chr.17: 68539423-68539423 on Build GRCh38
rs147490820 Chr.17: 68539357-68539357 on Build GRCh38
rs758703472 Chr.17: 68539330-68539331 on Build GRCh38
rs767668948 Chr.17: 68539434-68539434 on Build GRCh38
rs570497434 Chr.17: 68539393-68539393 on Build GRCh38
rs770683832 Chr.17: 68539300-68539300 on Build GRCh38
rs948224533 Chr.17: 68539443-68539443 on Build GRCh38
rs907879049 Chr.17: 68539426-68539426 on Build GRCh38
rs753713680 Chr.17: 68539328-68539328 on Build GRCh38
rs780415641 Chr.17: 68539388-68539388 on Build GRCh38
rs769233184 Chr.17: 68539373-68539373 on Build GRCh38
rs925588578 Chr.17: 68539469-68539469 on Build GRCh38
rs147636224 Chr.17: 68539383-68539383 on Build GRCh38
rs866926047 Chr.17: 68539347-68539347 on Build GRCh38
rs370186183 Chr.17: 68539311-68539311 on Build GRCh38
rs746449968 Chr.17: 68539369-68539369 on Build GRCh38
rs983444175 Chr.17: 68539420-68539420 on Build GRCh38
rs763069538 Chr.17: 68539401-68539401 on Build GRCh38
rs766545212 Chr.17: 68539307-68539307 on Build GRCh38
rs960813011 Chr.17: 68539428-68539428 on Build GRCh38
rs774510993 Chr.17: 68539429-68539429 on Build GRCh38
rs889468694 Chr.17: 68539364-68539364 on Build GRCh38
rs778825847 Chr.17: 68539355-68539355 on Build GRCh38
rs1052089651 Chr.17: 68539253-68539253 on Build GRCh38
rs952058870 Chr.17: 68539396-68539396 on Build GRCh38
rs773554835 Chr.17: 68539392-68539392 on Build GRCh38
rs78050433 Chr.17: 68539384-68539384 on Build GRCh38
rs139949522 Chr.17: 68539438-68539438 on Build GRCh38
rs763065277 Chr.17: 68539298-68539298 on Build GRCh38
rs376375756 Chr.17: 68539419-68539419 on Build GRCh38

View Full Product Details