Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200490464-200490693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD18
Entrez Gene ID
130535
Gene Name
potassium channel tetramerization domain containing 18
Gene Aliases
6530404F10Rik
UniGene
Hs.605775
Gene Chromosome Location
Chr.2: 200488952-200510094 on Build GRCh38
Transcripts
NM_001321547.1 NM_001321548.1 NM_001321550.1 NM_152387.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1683886 c.832delT p.L279fs*98 Chr.2: 200490549-200490549 on Build GRCh38
1014682 c.840C>T p.G280G Chr.2: 200490541-200490541 on Build GRCh38
2906375 c.836delT p.L279fs*98 Chr.2: 200490545-200490545 on Build GRCh38
2906374 c.899C>A p.A300D Chr.2: 200490482-200490482 on Build GRCh38
1014681 c.915G>A p.T305T Chr.2: 200490466-200490466 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374611164 Chr.2: 200490615-200490615 on Build GRCh38
rs747956684 Chr.2: 200490599-200490599 on Build GRCh38
rs368692549 Chr.2: 200490632-200490632 on Build GRCh38
rs200202417 Chr.2: 200490482-200490482 on Build GRCh38
rs748821162 Chr.2: 200490509-200490509 on Build GRCh38
rs534708442 Chr.2: 200490625-200490625 on Build GRCh38
rs775798899 Chr.2: 200490577-200490577 on Build GRCh38
rs369656416 Chr.2: 200490505-200490505 on Build GRCh38
rs769903059 Chr.2: 200490626-200490626 on Build GRCh38
rs149091190 Chr.2: 200490492-200490492 on Build GRCh38
rs763517829 Chr.2: 200490580-200490580 on Build GRCh38
rs771459770 Chr.2: 200490650-200490650 on Build GRCh38
rs754829938 Chr.2: 200490520-200490520 on Build GRCh38
rs200898789 Chr.2: 200490604-200490604 on Build GRCh38
rs922422319 Chr.2: 200490634-200490634 on Build GRCh38
rs1009532746 Chr.2: 200490549-200490549 on Build GRCh38
rs1002347034 Chr.2: 200490573-200490573 on Build GRCh38
rs578090485 Chr.2: 200490654-200490654 on Build GRCh38
rs780719949 Chr.2: 200490494-200490494 on Build GRCh38
rs115629407 Chr.2: 200490593-200490593 on Build GRCh38
rs748148780 Chr.2: 200490526-200490526 on Build GRCh38
rs370785664 Chr.2: 200490592-200490592 on Build GRCh38
rs778520508 Chr.2: 200490524-200490524 on Build GRCh38
rs776918225 Chr.2: 200490553-200490553 on Build GRCh38
rs372891645 Chr.2: 200490534-200490534 on Build GRCh38
rs765680919 Chr.2: 200490575-200490575 on Build GRCh38
rs376505965 Chr.2: 200490646-200490646 on Build GRCh38
rs370973122 Chr.2: 200490607-200490607 on Build GRCh38
rs569631375 Chr.2: 200490567-200490567 on Build GRCh38
rs776526393 Chr.2: 200490616-200490616 on Build GRCh38
rs201678106 Chr.2: 200490535-200490535 on Build GRCh38
rs770858936 Chr.2: 200490542-200490542 on Build GRCh38
rs181273720 Chr.2: 200490589-200490589 on Build GRCh38
rs774866286 Chr.2: 200490645-200490645 on Build GRCh38
rs368024909 Chr.2: 200490652-200490652 on Build GRCh38
rs758879282 Chr.2: 200490493-200490493 on Build GRCh38
rs764570627 Chr.2: 200490585-200490585 on Build GRCh38
rs376356569 Chr.2: 200490588-200490588 on Build GRCh38
rs765454221 Chr.2: 200490487-200490487 on Build GRCh38
rs200312807 Chr.2: 200490633-200490633 on Build GRCh38
rs777866408 Chr.2: 200490605-200490605 on Build GRCh38
rs752339754 Chr.2: 200490657-200490657 on Build GRCh38
rs866190955 Chr.2: 200490496-200490496 on Build GRCh38
rs765961264 Chr.2: 200490648-200490648 on Build GRCh38
rs188668776 Chr.2: 200490541-200490541 on Build GRCh38
rs370941047 Chr.2: 200490506-200490506 on Build GRCh38

View Full Product Details