Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28155865-28156106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELP3
Entrez Gene ID
55140
Gene Name
elongator acetyltransferase complex subunit 3
Gene Aliases
KAT9
UniGene
Hs.491336
Gene Chromosome Location
Chr.8: 28090231-28191153 on Build GRCh38
Transcripts
NM_001284220.1 NM_001284222.1 NM_001284224.1 NM_001284225.1 NM_001284226.1 NM_018091.5 XM_006716354.2 XM_017013604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5773756 c.1119A>C p.L373F Chr.8: 28155960-28155960 on Build GRCh38
1191041 c.1142_1143insAGTGG p.N382fs*4 Chr.8: 28155983-28155984 on Build GRCh38
6231834 c.1101-3C>A p.? Chr.8: 28155939-28155939 on Build GRCh38
4922287 c.1112T>C p.M371T Chr.8: 28155953-28155953 on Build GRCh38
1098854 c.1106T>G p.I369S Chr.8: 28155947-28155947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373841521 Chr.8: 28156024-28156024 on Build GRCh38
rs745496625 Chr.8: 28156051-28156051 on Build GRCh38
rs199903018 Chr.8: 28155952-28155952 on Build GRCh38
rs770553945 Chr.8: 28155894-28155894 on Build GRCh38
rs748769151 Chr.8: 28156049-28156049 on Build GRCh38
rs750706913 Chr.8: 28156001-28156001 on Build GRCh38
rs190950615 Chr.8: 28156050-28156050 on Build GRCh38
rs775866720 Chr.8: 28156064-28156064 on Build GRCh38
rs908496437 Chr.8: 28156054-28156054 on Build GRCh38
rs761177498 Chr.8: 28155935-28155935 on Build GRCh38
rs1033280697 Chr.8: 28156043-28156043 on Build GRCh38
rs761004441 Chr.8: 28156068-28156068 on Build GRCh38
rs746560423 Chr.8: 28155985-28155985 on Build GRCh38
rs143728730 Chr.8: 28156049-28156050 on Build GRCh38
rs749370251 Chr.8: 28155955-28155955 on Build GRCh38
rs755055081 Chr.8: 28155974-28155975 on Build GRCh38
rs1021985398 Chr.8: 28155994-28155994 on Build GRCh38
rs746877030 Chr.8: 28156001-28156001 on Build GRCh38
rs780997433 Chr.8: 28156008-28156008 on Build GRCh38
rs769283470 Chr.8: 28156081-28156081 on Build GRCh38
rs142534142 Chr.8: 28155921-28155921 on Build GRCh38
rs180852092 Chr.8: 28155895-28155895 on Build GRCh38
rs141713695 Chr.8: 28155961-28155961 on Build GRCh38
rs868372601 Chr.8: 28155982-28155983 on Build GRCh38
rs752434486 Chr.8: 28156023-28156023 on Build GRCh38
rs959181955 Chr.8: 28155980-28155980 on Build GRCh38
rs777742193 Chr.8: 28156036-28156036 on Build GRCh38
rs4732832 Chr.8: 28156062-28156062 on Build GRCh38
rs759802141 Chr.8: 28155918-28155918 on Build GRCh38
rs777767559 Chr.8: 28155983-28155984 on Build GRCh38
rs765523825 Chr.8: 28155993-28155993 on Build GRCh38
rs764713897 Chr.8: 28155948-28155948 on Build GRCh38
rs780350740 Chr.8: 28155912-28155912 on Build GRCh38
rs370302582 Chr.8: 28156007-28156007 on Build GRCh38
rs777316486 Chr.8: 28156082-28156082 on Build GRCh38
rs892624094 Chr.8: 28155924-28155924 on Build GRCh38

View Full Product Details