Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 13496003-13496254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS3ST3A1
Entrez Gene ID
9955
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 3A1
Gene Aliases
3-OST-3A, 3OST3A1
UniGene
Hs.462270
Gene Chromosome Location
Chr.17: 13494032-13601942 on Build GRCh38
Transcripts
NM_006042.2 XM_011524114.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3514414 c.1178A>C p.K393T Chr.17: 13496240-13496240 on Build GRCh38
7101807 c.1179G>C p.K393N Chr.17: 13496239-13496239 on Build GRCh38
3514413 c.1188G>A p.Q396Q Chr.17: 13496230-13496230 on Build GRCh38
6682230 c.1214A>G p.D405G Chr.17: 13496204-13496204 on Build GRCh38
6682229 c.1211G>T p.W404L Chr.17: 13496207-13496207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958996872 Chr.17: 13496079-13496079 on Build GRCh38
rs977530011 Chr.17: 13496140-13496140 on Build GRCh38
rs910341683 Chr.17: 13496207-13496207 on Build GRCh38
rs772621059 Chr.17: 13496210-13496210 on Build GRCh38
rs958855086 Chr.17: 13496099-13496099 on Build GRCh38
rs780830063 Chr.17: 13496186-13496186 on Build GRCh38
rs370717555 Chr.17: 13496223-13496223 on Build GRCh38
rs746415528 Chr.17: 13496205-13496205 on Build GRCh38
rs923717856 Chr.17: 13496226-13496226 on Build GRCh38
rs67951062 Chr.17: 13496170-13496171 on Build GRCh38
rs765869598 Chr.17: 13496172-13496172 on Build GRCh38
rs527418205 Chr.17: 13496083-13496083 on Build GRCh38
rs762966288 Chr.17: 13496168-13496169 on Build GRCh38
rs538062564 Chr.17: 13496170-13496170 on Build GRCh38
rs984307026 Chr.17: 13496059-13496059 on Build GRCh38
rs779396044 Chr.17: 13496201-13496201 on Build GRCh38
rs567189152 Chr.17: 13496102-13496102 on Build GRCh38
rs761556942 Chr.17: 13496155-13496155 on Build GRCh38
rs993285873 Chr.17: 13496029-13496029 on Build GRCh38
rs878977677 Chr.17: 13496152-13496152 on Build GRCh38
rs879522134 Chr.17: 13496179-13496180 on Build GRCh38
rs998579735 Chr.17: 13496049-13496049 on Build GRCh38
rs901197622 Chr.17: 13496032-13496032 on Build GRCh38
rs560355812 Chr.17: 13496055-13496055 on Build GRCh38
rs1035784162 Chr.17: 13496077-13496077 on Build GRCh38
rs970422812 Chr.17: 13496073-13496074 on Build GRCh38
rs750411039 Chr.17: 13496167-13496167 on Build GRCh38
rs1003016898 Chr.17: 13496073-13496073 on Build GRCh38
rs775448387 Chr.17: 13496217-13496217 on Build GRCh38
rs776298980 Chr.17: 13496146-13496146 on Build GRCh38
rs770247855 Chr.17: 13496147-13496148 on Build GRCh38
rs766707176 Chr.17: 13496169-13496170 on Build GRCh38
rs751065391 Chr.17: 13496173-13496173 on Build GRCh38
rs537843894 Chr.17: 13496143-13496143 on Build GRCh38
rs190268276 Chr.17: 13496047-13496047 on Build GRCh38
rs202026672 Chr.17: 13496221-13496221 on Build GRCh38
rs11385090 Chr.17: 13496178-13496179 on Build GRCh38
rs754599728 Chr.17: 13496180-13496180 on Build GRCh38
rs371396064 Chr.17: 13496196-13496196 on Build GRCh38
rs755906914 Chr.17: 13496175-13496176 on Build GRCh38
rs969103774 Chr.17: 13496108-13496108 on Build GRCh38
rs776785481 Chr.17: 13496224-13496224 on Build GRCh38
rs773729992 Chr.17: 13496155-13496159 on Build GRCh38
rs549042711 Chr.17: 13496095-13496095 on Build GRCh38
rs1025739953 Chr.17: 13496054-13496054 on Build GRCh38
rs749696540 Chr.17: 13496187-13496187 on Build GRCh38
rs765182611 Chr.17: 13496162-13496162 on Build GRCh38
rs565296202 Chr.17: 13496218-13496218 on Build GRCh38
rs758301806 Chr.17: 13496169-13496169 on Build GRCh38

View Full Product Details