Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 136928079-136928304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCK1
Entrez Gene ID
4690
Gene Name
NCK adaptor protein 1
Gene Aliases
NCK, NCKalpha, nck-1
UniGene
Hs.126889
Gene Chromosome Location
Chr.3: 136862208-136951610 on Build GRCh38
Transcripts
NM_001291999.1 NM_006153.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
728221 c.118C>T p.R40* Chr.3: 136928119-136928119 on Build GRCh38
5465725 c.220A>G p.T74A Chr.3: 136928221-136928221 on Build GRCh38
3123490 c.149T>C p.F50S Chr.3: 136928150-136928150 on Build GRCh38
84829 c.187C>T p.R63W Chr.3: 136928188-136928188 on Build GRCh38
6205071 c.188G>A p.R63Q Chr.3: 136928189-136928189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763704002 Chr.3: 136928163-136928163 on Build GRCh38
rs746401849 Chr.3: 136928258-136928258 on Build GRCh38
rs774656852 Chr.3: 136928251-136928251 on Build GRCh38
rs889691574 Chr.3: 136928242-136928242 on Build GRCh38
rs370617853 Chr.3: 136928203-136928203 on Build GRCh38
rs763295518 Chr.3: 136928240-136928241 on Build GRCh38
rs772639807 Chr.3: 136928166-136928166 on Build GRCh38
rs771394511 Chr.3: 136928152-136928152 on Build GRCh38
rs537275699 Chr.3: 136928189-136928189 on Build GRCh38
rs749678194 Chr.3: 136928235-136928235 on Build GRCh38
rs768235189 Chr.3: 136928259-136928259 on Build GRCh38
rs748380862 Chr.3: 136928111-136928111 on Build GRCh38
rs201444058 Chr.3: 136928223-136928223 on Build GRCh38
rs765039223 Chr.3: 136928271-136928271 on Build GRCh38
rs778198006 Chr.3: 136928119-136928119 on Build GRCh38
rs761527835 Chr.3: 136928266-136928266 on Build GRCh38
rs771468274 Chr.3: 136928240-136928240 on Build GRCh38
rs748325789 Chr.3: 136928214-136928214 on Build GRCh38
rs770040215 Chr.3: 136928218-136928218 on Build GRCh38
rs773593965 Chr.3: 136928120-136928120 on Build GRCh38
rs763481977 Chr.3: 136928148-136928148 on Build GRCh38
rs1028209928 Chr.3: 136928241-136928242 on Build GRCh38
rs751517125 Chr.3: 136928196-136928196 on Build GRCh38
rs779922033 Chr.3: 136928191-136928191 on Build GRCh38
rs771451936 Chr.3: 136928200-136928200 on Build GRCh38
rs760172881 Chr.3: 136928160-136928160 on Build GRCh38
rs761581267 Chr.3: 136928182-136928182 on Build GRCh38
rs1008264920 Chr.3: 136928246-136928246 on Build GRCh38
rs148772850 Chr.3: 136928186-136928186 on Build GRCh38
rs200674609 Chr.3: 136928263-136928263 on Build GRCh38
rs750918907 Chr.3: 136928235-136928235 on Build GRCh38
rs775056529 Chr.3: 136928159-136928159 on Build GRCh38
rs898267362 Chr.3: 136928217-136928217 on Build GRCh38

View Full Product Details