Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 96246980-96247227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELK3
Entrez Gene ID
2004
Gene Name
ELK3, ETS transcription factor
Gene Aliases
ERP, NET, SAP-2, SAP2
UniGene
Hs.46523
Gene Chromosome Location
Chr.12: 96194382-96269835 on Build GRCh38
Transcripts
NM_005230.3 XM_006719275.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
118450 c.336C>T p.C112C Chr.12: 96247068-96247068 on Build GRCh38
3466661 c.261C>T p.F87F Chr.12: 96246993-96246993 on Build GRCh38
7194432 c.433T>G p.S145A Chr.12: 96247165-96247165 on Build GRCh38
3417299 c.263C>T p.P88L Chr.12: 96246995-96246995 on Build GRCh38
7113601 c.395C>T p.T132M Chr.12: 96247127-96247127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770276677 Chr.12: 96247074-96247074 on Build GRCh38
rs754714305 Chr.12: 96247167-96247167 on Build GRCh38
rs372868841 Chr.12: 96247020-96247020 on Build GRCh38
rs771444556 Chr.12: 96247132-96247132 on Build GRCh38
rs1043041666 Chr.12: 96247060-96247060 on Build GRCh38
rs773841437 Chr.12: 96247076-96247076 on Build GRCh38
rs878882439 Chr.12: 96247192-96247192 on Build GRCh38
rs546312459 Chr.12: 96247090-96247090 on Build GRCh38
rs778689224 Chr.12: 96247036-96247036 on Build GRCh38
rs1063999 Chr.12: 96247186-96247186 on Build GRCh38
rs112218128 Chr.12: 96247128-96247128 on Build GRCh38
rs1063997 Chr.12: 96247073-96247073 on Build GRCh38
rs755735999 Chr.12: 96247194-96247194 on Build GRCh38
rs778883490 Chr.12: 96247100-96247100 on Build GRCh38
rs371589438 Chr.12: 96247063-96247063 on Build GRCh38
rs140037312 Chr.12: 96247079-96247079 on Build GRCh38
rs763626966 Chr.12: 96247016-96247016 on Build GRCh38
rs747750273 Chr.12: 96247127-96247127 on Build GRCh38
rs202181482 Chr.12: 96247114-96247114 on Build GRCh38
rs201510944 Chr.12: 96247200-96247200 on Build GRCh38
rs368919889 Chr.12: 96247201-96247201 on Build GRCh38
rs751829052 Chr.12: 96247025-96247025 on Build GRCh38
rs747726707 Chr.12: 96247181-96247181 on Build GRCh38
rs752654847 Chr.12: 96247088-96247088 on Build GRCh38
rs1033592762 Chr.12: 96247095-96247095 on Build GRCh38
rs755054684 Chr.12: 96247027-96247027 on Build GRCh38
rs142269306 Chr.12: 96247104-96247104 on Build GRCh38
rs374221949 Chr.12: 96247002-96247002 on Build GRCh38
rs776166918 Chr.12: 96247065-96247065 on Build GRCh38
rs750380483 Chr.12: 96247158-96247158 on Build GRCh38
rs763534472 Chr.12: 96247130-96247130 on Build GRCh38
rs767724593 Chr.12: 96247148-96247148 on Build GRCh38
rs764133272 Chr.12: 96247155-96247155 on Build GRCh38
rs746668569 Chr.12: 96247112-96247112 on Build GRCh38
rs760878221 Chr.12: 96247149-96247149 on Build GRCh38
rs368663242 Chr.12: 96247062-96247062 on Build GRCh38
rs766764247 Chr.12: 96247024-96247024 on Build GRCh38
rs756305673 Chr.12: 96247113-96247113 on Build GRCh38
rs377633718 Chr.12: 96247164-96247164 on Build GRCh38
rs767528787 Chr.12: 96247089-96247089 on Build GRCh38
rs779718219 Chr.12: 96247042-96247042 on Build GRCh38
rs758289104 Chr.12: 96247159-96247159 on Build GRCh38
rs200818787 Chr.12: 96247023-96247023 on Build GRCh38
rs760242678 Chr.12: 96247005-96247005 on Build GRCh38
rs142263510 Chr.12: 96247137-96247137 on Build GRCh38
rs1063998 Chr.12: 96247115-96247115 on Build GRCh38
rs765550323 Chr.12: 96247022-96247022 on Build GRCh38
rs1025986001 Chr.12: 96247185-96247185 on Build GRCh38
rs758236190 Chr.12: 96247105-96247105 on Build GRCh38
rs560926679 Chr.12: 96247029-96247029 on Build GRCh38
rs375568901 Chr.12: 96247021-96247021 on Build GRCh38
rs745607623 Chr.12: 96247037-96247037 on Build GRCh38
rs762230733 Chr.12: 96247067-96247067 on Build GRCh38
rs751879824 Chr.12: 96247087-96247087 on Build GRCh38
rs779828379 Chr.12: 96247163-96247163 on Build GRCh38
rs757158344 Chr.12: 96247092-96247092 on Build GRCh38
rs759686205 Chr.12: 96247138-96247138 on Build GRCh38
rs185083482 Chr.12: 96247080-96247080 on Build GRCh38
rs780963033 Chr.12: 96247172-96247172 on Build GRCh38
rs757182383 Chr.12: 96247031-96247031 on Build GRCh38
rs910759899 Chr.12: 96247197-96247197 on Build GRCh38

View Full Product Details