Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 127582912-127583163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCC1
Entrez Gene ID
79571
Gene Name
GRIP and coiled-coil domain containing 1
Gene Aliases
GCC1P, GCC88
UniGene
Hs.521168
Gene Chromosome Location
Chr.7: 127580628-127585600 on Build GRCh38
Transcripts
NM_024523.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1085143 c.1190G>A p.R397H Chr.7: 127583152-127583152 on Build GRCh38
4825228 c.1242C>A p.S414R Chr.7: 127583100-127583100 on Build GRCh38
395363 c.1395G>T p.M465I Chr.7: 127582947-127582947 on Build GRCh38
3784052 c.1336G>A p.A446T Chr.7: 127583006-127583006 on Build GRCh38
244672 c.1386G>T p.L462L Chr.7: 127582956-127582956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55695269 Chr.7: 127583053-127583053 on Build GRCh38
rs772605757 Chr.7: 127583009-127583009 on Build GRCh38
rs753969534 Chr.7: 127583087-127583087 on Build GRCh38
rs767361349 Chr.7: 127583060-127583060 on Build GRCh38
rs759319387 Chr.7: 127583026-127583026 on Build GRCh38
rs374662823 Chr.7: 127583085-127583085 on Build GRCh38
rs111491677 Chr.7: 127582960-127582960 on Build GRCh38
rs762751259 Chr.7: 127583038-127583038 on Build GRCh38
rs758362532 Chr.7: 127582967-127582967 on Build GRCh38
rs200740651 Chr.7: 127583004-127583004 on Build GRCh38
rs368859467 Chr.7: 127582946-127582946 on Build GRCh38
rs778288314 Chr.7: 127583121-127583121 on Build GRCh38
rs756138761 Chr.7: 127582990-127582990 on Build GRCh38
rs999001094 Chr.7: 127583051-127583051 on Build GRCh38
rs781365548 Chr.7: 127582983-127582983 on Build GRCh38
rs776417081 Chr.7: 127583017-127583017 on Build GRCh38
rs772550692 Chr.7: 127583113-127583113 on Build GRCh38
rs774780646 Chr.7: 127582938-127582938 on Build GRCh38
rs775099457 Chr.7: 127583035-127583035 on Build GRCh38
rs763757405 Chr.7: 127582973-127582973 on Build GRCh38
rs751379461 Chr.7: 127582982-127582982 on Build GRCh38
rs903430442 Chr.7: 127583114-127583114 on Build GRCh38
rs202171274 Chr.7: 127583108-127583108 on Build GRCh38
rs775055157 Chr.7: 127583135-127583135 on Build GRCh38
rs142791828 Chr.7: 127583127-127583127 on Build GRCh38
rs750588727 Chr.7: 127583067-127583067 on Build GRCh38
rs199928954 Chr.7: 127582959-127582959 on Build GRCh38
rs756361154 Chr.7: 127583079-127583079 on Build GRCh38
rs199528674 Chr.7: 127583007-127583007 on Build GRCh38
rs760571264 Chr.7: 127582947-127582947 on Build GRCh38
rs745472409 Chr.7: 127583126-127583126 on Build GRCh38
rs751541233 Chr.7: 127583045-127583045 on Build GRCh38
rs769509250 Chr.7: 127583129-127583129 on Build GRCh38
rs764071715 Chr.7: 127583040-127583040 on Build GRCh38
rs894573216 Chr.7: 127583041-127583041 on Build GRCh38
rs764868196 Chr.7: 127582952-127582952 on Build GRCh38
rs903386948 Chr.7: 127583046-127583046 on Build GRCh38
rs34836950 Chr.7: 127583028-127583028 on Build GRCh38
rs982002535 Chr.7: 127582984-127582984 on Build GRCh38
rs746158991 Chr.7: 127582989-127582989 on Build GRCh38
rs371600897 Chr.7: 127582942-127582942 on Build GRCh38
rs748729535 Chr.7: 127583109-127583109 on Build GRCh38
rs755058694 Chr.7: 127583094-127583094 on Build GRCh38
rs890689823 Chr.7: 127583056-127583056 on Build GRCh38
rs749688639 Chr.7: 127583000-127583000 on Build GRCh38
rs762497377 Chr.7: 127583141-127583141 on Build GRCh38
rs951298265 Chr.7: 127583120-127583120 on Build GRCh38
rs140613801 Chr.7: 127582992-127582992 on Build GRCh38
rs766151079 Chr.7: 127582941-127582941 on Build GRCh38
rs760360160 Chr.7: 127582939-127582939 on Build GRCh38
rs774525979 Chr.7: 127583005-127583005 on Build GRCh38

View Full Product Details