Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3359062-3359335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF195
Entrez Gene ID
7748
Gene Name
zinc finger protein 195
Gene Aliases
HRF1, ZNFP104
UniGene
Hs.386294
Gene Chromosome Location
Chr.11: 3357927-3379222 on Build GRCh38
Transcripts
NM_001130519.2 NM_001130520.2 NM_001242841.1 NM_001242842.1 NM_001242843.1 NM_001256823.1 NM_001256824.1 NM_001256825.1 NM_007152.4 NR_040083.1 NR_046381.1 NR_046382.1 XM_006718306.1 XM_011520350.2 XM_011520351.2 XM_011520352.2 XM_017018260.1 XM_017018261.1 XM_017018262.1 XM_017018263.1 XM_017018264.1 XM_017018265.1 XM_017018266.1 XM_017018267.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
926564 c.1528G>A p.E510K Chr.11: 3359264-3359264 on Build GRCh38
3869105 c.1728A>G p.K576K Chr.11: 3359280-3359280 on Build GRCh38
6449826 c.1830G>A p.K610K Chr.11: 3359178-3359178 on Build GRCh38
6828330 c.1764G>A p.K588K Chr.11: 3359187-3359187 on Build GRCh38
3979591 c.1601A>G p.Y534C Chr.11: 3359191-3359191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376866898 Chr.11: 3359229-3359229 on Build GRCh38
rs772652328 Chr.11: 3359204-3359204 on Build GRCh38
rs371443966 Chr.11: 3359116-3359116 on Build GRCh38
rs762194270 Chr.11: 3359241-3359241 on Build GRCh38
rs779876785 Chr.11: 3359264-3359264 on Build GRCh38
rs369261399 Chr.11: 3359083-3359083 on Build GRCh38
rs767767271 Chr.11: 3359247-3359247 on Build GRCh38
rs750520434 Chr.11: 3359250-3359250 on Build GRCh38
rs551670550 Chr.11: 3359212-3359212 on Build GRCh38
rs776102952 Chr.11: 3359226-3359226 on Build GRCh38
rs748694606 Chr.11: 3359153-3359153 on Build GRCh38
rs749111666 Chr.11: 3359166-3359166 on Build GRCh38
rs1033189679 Chr.11: 3359206-3359206 on Build GRCh38
rs933818026 Chr.11: 3359305-3359305 on Build GRCh38
rs771748449 Chr.11: 3359194-3359194 on Build GRCh38
rs754771154 Chr.11: 3359308-3359308 on Build GRCh38
rs759607316 Chr.11: 3359152-3359152 on Build GRCh38
rs957567673 Chr.11: 3359220-3359220 on Build GRCh38
rs780019712 Chr.11: 3359145-3359145 on Build GRCh38
rs764499611 Chr.11: 3359232-3359232 on Build GRCh38
rs922407176 Chr.11: 3359280-3359280 on Build GRCh38
rs763436510 Chr.11: 3359231-3359231 on Build GRCh38
rs768390052 Chr.11: 3359181-3359181 on Build GRCh38
rs763381090 Chr.11: 3359099-3359099 on Build GRCh38
rs756189578 Chr.11: 3359262-3359262 on Build GRCh38
rs777187713 Chr.11: 3359225-3359225 on Build GRCh38
rs779493378 Chr.11: 3359109-3359110 on Build GRCh38
rs753716888 Chr.11: 3359265-3359265 on Build GRCh38
rs751839475 Chr.11: 3359235-3359235 on Build GRCh38
rs753049436 Chr.11: 3359092-3359092 on Build GRCh38
rs756066763 Chr.11: 3359142-3359142 on Build GRCh38
rs755661378 Chr.11: 3359110-3359110 on Build GRCh38
rs747732338 Chr.11: 3359191-3359191 on Build GRCh38
rs750511570 Chr.11: 3359131-3359131 on Build GRCh38
rs146782590 Chr.11: 3359113-3359113 on Build GRCh38
rs772513846 Chr.11: 3359250-3359251 on Build GRCh38
rs760208512 Chr.11: 3359205-3359205 on Build GRCh38
rs764444371 Chr.11: 3359105-3359105 on Build GRCh38
rs778550359 Chr.11: 3359185-3359185 on Build GRCh38
rs757428990 Chr.11: 3359122-3359122 on Build GRCh38
rs778257565 Chr.11: 3359274-3359274 on Build GRCh38
rs781066718 Chr.11: 3359123-3359123 on Build GRCh38
rs141992358 Chr.11: 3359102-3359105 on Build GRCh38

View Full Product Details