Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143817297-143817522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PUF60
Entrez Gene ID
22827
Gene Name
poly(U) binding splicing factor 60
Gene Aliases
FIR, RoBPI, SIAHBP1, VRJS
UniGene
Hs.521924
Gene Chromosome Location
Chr.8: 143816344-143829386 on Build GRCh38
Transcripts
NM_001136033.2 NM_001271096.1 NM_001271097.1 NM_001271098.1 NM_001271099.1 NM_001271100.1 NM_014281.4 NM_078480.2 XM_011516929.2 XM_011516930.2 XM_017013234.1 XM_017013235.1 XM_017013236.1 XM_017013237.1 XM_017013238.1 XM_017013239.1 XM_017013240.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2868894 c.1101G>T p.L367F Chr.8: 143817374-143817374 on Build GRCh38
1097046 c.1028C>T p.A343V Chr.8: 143817447-143817447 on Build GRCh38
6230942 c.1127C>T p.A376V Chr.8: 143817348-143817348 on Build GRCh38
4608010 c.1120G>T p.A374S Chr.8: 143817355-143817355 on Build GRCh38
7424899 c.1043T>C p.L348P Chr.8: 143817432-143817432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775532345 Chr.8: 143817487-143817487 on Build GRCh38
rs368804058 Chr.8: 143817446-143817446 on Build GRCh38
rs749620335 Chr.8: 143817428-143817428 on Build GRCh38
rs756595524 Chr.8: 143817413-143817413 on Build GRCh38
rs758312311 Chr.8: 143817340-143817340 on Build GRCh38
rs374716646 Chr.8: 143817486-143817486 on Build GRCh38
rs1024946487 Chr.8: 143817403-143817403 on Build GRCh38
rs768969026 Chr.8: 143817433-143817433 on Build GRCh38
rs372391932 Chr.8: 143817482-143817482 on Build GRCh38
rs751319236 Chr.8: 143817494-143817494 on Build GRCh38
rs535427378 Chr.8: 143817335-143817335 on Build GRCh38
rs539674690 Chr.8: 143817489-143817489 on Build GRCh38
rs1002163574 Chr.8: 143817397-143817397 on Build GRCh38
rs772250893 Chr.8: 143817469-143817469 on Build GRCh38
rs1014103950 Chr.8: 143817385-143817385 on Build GRCh38
rs751881683 Chr.8: 143817434-143817434 on Build GRCh38
rs200407902 Chr.8: 143817490-143817490 on Build GRCh38
rs35881230 Chr.8: 143817368-143817368 on Build GRCh38
rs780683084 Chr.8: 143817404-143817404 on Build GRCh38
rs773329778 Chr.8: 143817477-143817477 on Build GRCh38
rs201956893 Chr.8: 143817424-143817424 on Build GRCh38
rs752519563 Chr.8: 143817328-143817328 on Build GRCh38
rs764016887 Chr.8: 143817376-143817376 on Build GRCh38
rs200345275 Chr.8: 143817380-143817380 on Build GRCh38
rs374293328 Chr.8: 143817418-143817418 on Build GRCh38
rs748341904 Chr.8: 143817455-143817455 on Build GRCh38
rs751177685 Chr.8: 143817378-143817378 on Build GRCh38
rs761518973 Chr.8: 143817501-143817501 on Build GRCh38
rs371162365 Chr.8: 143817410-143817410 on Build GRCh38

View Full Product Details