Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67304912-67305147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSH3
Entrez Gene ID
54961
Gene Name
slingshot protein phosphatase 3
Gene Aliases
SSH3L
UniGene
Hs.29173
Gene Chromosome Location
Chr.11: 67303448-67312607 on Build GRCh38
Transcripts
NM_017857.3 XM_017017942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6212802 c.271C>T p.Q91* Chr.11: 67304939-67304939 on Build GRCh38
467302 c.244G>A p.G82R Chr.11: 67304912-67304912 on Build GRCh38
3931418 c.254C>T p.S85F Chr.11: 67304922-67304922 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs537861551 Chr.11: 67305069-67305069 on Build GRCh38
rs763159223 Chr.11: 67305015-67305015 on Build GRCh38
rs146605334 Chr.11: 67304947-67304947 on Build GRCh38
rs370738715 Chr.11: 67304965-67304965 on Build GRCh38
rs770337915 Chr.11: 67305041-67305041 on Build GRCh38
rs767557682 Chr.11: 67305026-67305026 on Build GRCh38
rs150042639 Chr.11: 67304941-67304941 on Build GRCh38
rs1028983443 Chr.11: 67305117-67305117 on Build GRCh38
rs766030885 Chr.11: 67305084-67305084 on Build GRCh38
rs890129839 Chr.11: 67305004-67305004 on Build GRCh38
rs767972468 Chr.11: 67304934-67304945 on Build GRCh38
rs746427282 Chr.11: 67305040-67305040 on Build GRCh38
rs752542989 Chr.11: 67305032-67305032 on Build GRCh38
rs369895961 Chr.11: 67305030-67305030 on Build GRCh38
rs377583512 Chr.11: 67304960-67304960 on Build GRCh38
rs567204945 Chr.11: 67305103-67305103 on Build GRCh38
rs766601407 Chr.11: 67305030-67305030 on Build GRCh38
rs754773644 Chr.11: 67304970-67304970 on Build GRCh38
rs746534042 Chr.11: 67304997-67304997 on Build GRCh38
rs530739672 Chr.11: 67305003-67305003 on Build GRCh38
rs770114690 Chr.11: 67305008-67305008 on Build GRCh38
rs1056219076 Chr.11: 67304975-67304975 on Build GRCh38
rs141467579 Chr.11: 67304948-67304948 on Build GRCh38
rs753825437 Chr.11: 67305029-67305029 on Build GRCh38
rs780363712 Chr.11: 67305051-67305051 on Build GRCh38
rs777590448 Chr.11: 67305034-67305034 on Build GRCh38
rs778751731 Chr.11: 67304983-67304983 on Build GRCh38
rs143908566 Chr.11: 67305002-67305002 on Build GRCh38
rs941903504 Chr.11: 67304939-67304939 on Build GRCh38
rs566196481 Chr.11: 67305011-67305011 on Build GRCh38
rs777502459 Chr.11: 67304989-67304989 on Build GRCh38
rs757267293 Chr.11: 67304962-67304962 on Build GRCh38
rs896207008 Chr.11: 67304976-67304976 on Build GRCh38
rs774504600 Chr.11: 67305020-67305020 on Build GRCh38
rs766322478 Chr.11: 67304963-67304963 on Build GRCh38
rs1008440116 Chr.11: 67305035-67305035 on Build GRCh38
rs773435339 Chr.11: 67305029-67305030 on Build GRCh38
rs764387579 Chr.11: 67305017-67305017 on Build GRCh38
rs761908731 Chr.11: 67305024-67305024 on Build GRCh38
rs558259386 Chr.11: 67305038-67305038 on Build GRCh38
rs182079619 Chr.11: 67304952-67304952 on Build GRCh38
rs373641394 Chr.11: 67304985-67304985 on Build GRCh38
rs758068169 Chr.11: 67304988-67304988 on Build GRCh38
rs751217589 Chr.11: 67305037-67305037 on Build GRCh38
rs758073449 Chr.11: 67305033-67305033 on Build GRCh38
rs200871845 Chr.11: 67305052-67305052 on Build GRCh38

View Full Product Details