Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16465571-16465796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOSTDC1
Entrez Gene ID
25928
Gene Name
sclerostin domain containing 1
Gene Aliases
CDA019, DAND7, ECTODIN, USAG1
UniGene
Hs.648106
Gene Chromosome Location
Chr.7: 16461481-16496513 on Build GRCh38
Transcripts
NM_015464.2 XM_017011943.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3879934 c.94C>A p.L32I Chr.7: 16465575-16465575 on Build GRCh38
1329916 c.34C>G p.P12A Chr.7: 16465635-16465635 on Build GRCh38
72763 c.70T>C p.F24L Chr.7: 16465599-16465599 on Build GRCh38
4600578 c.28C>T p.L10F Chr.7: 16465641-16465641 on Build GRCh38
5744912 c.77delA p.N26fs*25 Chr.7: 16465592-16465592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749975640 Chr.7: 16465656-16465656 on Build GRCh38
rs778769557 Chr.7: 16465696-16465696 on Build GRCh38
rs899499404 Chr.7: 16465729-16465729 on Build GRCh38
rs1044358021 Chr.7: 16465752-16465752 on Build GRCh38
rs1008352096 Chr.7: 16465684-16465684 on Build GRCh38
rs748143306 Chr.7: 16465701-16465701 on Build GRCh38
rs779523341 Chr.7: 16465670-16465670 on Build GRCh38
rs768545948 Chr.7: 16465686-16465686 on Build GRCh38
rs771727563 Chr.7: 16465705-16465705 on Build GRCh38
rs952711477 Chr.7: 16465767-16465767 on Build GRCh38
rs146877090 Chr.7: 16465655-16465655 on Build GRCh38
rs757515286 Chr.7: 16465649-16465649 on Build GRCh38
rs755994094 Chr.7: 16465668-16465668 on Build GRCh38
rs996756668 Chr.7: 16465750-16465750 on Build GRCh38
rs1042249101 Chr.7: 16465664-16465664 on Build GRCh38
rs544726803 Chr.7: 16465657-16465657 on Build GRCh38
rs201141429 Chr.7: 16465643-16465643 on Build GRCh38
rs780117735 Chr.7: 16465675-16465676 on Build GRCh38
rs764764953 Chr.7: 16465613-16465613 on Build GRCh38
rs531608232 Chr.7: 16465775-16465775 on Build GRCh38
rs374033488 Chr.7: 16465679-16465679 on Build GRCh38
rs6945425 Chr.7: 16465716-16465716 on Build GRCh38
rs750191548 Chr.7: 16465615-16465615 on Build GRCh38
rs867509981 Chr.7: 16465615-16465615 on Build GRCh38
rs567806396 Chr.7: 16465772-16465772 on Build GRCh38
rs527371758 Chr.7: 16465754-16465754 on Build GRCh38
rs746302000 Chr.7: 16465755-16465755 on Build GRCh38

View Full Product Details