Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 100584896-100585155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNMD
Entrez Gene ID
64102
Gene Name
tenomodulin
Gene Aliases
BRICD4, CHM1L, TEM
UniGene
Hs.132957
Gene Chromosome Location
Chr.X: 100584793-100599885 on Build GRCh38
Transcripts
NM_022144.2

SNP Detail

SNP ID Chromosome Location
rs369353974 Chr.X: 100585022-100585022 on Build GRCh38
rs748819768 Chr.X: 100585058-100585058 on Build GRCh38
rs760194821 Chr.X: 100585081-100585082 on Build GRCh38
rs753122283 Chr.X: 100584988-100584988 on Build GRCh38
rs771947367 Chr.X: 100585097-100585097 on Build GRCh38
rs549181515 Chr.X: 100584962-100584962 on Build GRCh38
rs759906894 Chr.X: 100584978-100584978 on Build GRCh38
rs201621778 Chr.X: 100585015-100585015 on Build GRCh38
rs199869003 Chr.X: 100585059-100585059 on Build GRCh38
rs758728379 Chr.X: 100585007-100585007 on Build GRCh38
rs202136339 Chr.X: 100585016-100585016 on Build GRCh38
rs950194589 Chr.X: 100585118-100585118 on Build GRCh38
rs761522235 Chr.X: 100585033-100585033 on Build GRCh38
rs200406916 Chr.X: 100585105-100585105 on Build GRCh38
rs765756778 Chr.X: 100584984-100584984 on Build GRCh38
rs769282025 Chr.X: 100585060-100585060 on Build GRCh38
rs376008251 Chr.X: 100584976-100584976 on Build GRCh38
rs779363146 Chr.X: 100585044-100585044 on Build GRCh38
rs36082211 Chr.X: 100585042-100585042 on Build GRCh38
rs760055425 Chr.X: 100585106-100585106 on Build GRCh38
rs955228991 Chr.X: 100584940-100584940 on Build GRCh38
rs748006128 Chr.X: 100585091-100585091 on Build GRCh38
rs942781194 Chr.X: 100584997-100584997 on Build GRCh38
rs1028595938 Chr.X: 100584939-100584939 on Build GRCh38
rs1008141235 Chr.X: 100584959-100584959 on Build GRCh38

View Full Product Details