Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46687009-46687264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TP53RK
Entrez Gene ID
112858
Gene Name
TP53 regulating kinase
Gene Aliases
BUD32, C20orf64, Nori-2, Nori-2p, PRPK, dJ101A2
UniGene
Hs.440263
Gene Chromosome Location
Chr.20: 46684365-46689637 on Build GRCh38
Transcripts
NM_033550.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3379068 c.396T>C p.T132T Chr.20: 46687119-46687119 on Build GRCh38
3423695 c.455G>A p.R152Q Chr.20: 46687060-46687060 on Build GRCh38
4944706 c.497C>T p.S166F Chr.20: 46687018-46687018 on Build GRCh38
4099042 c.440G>A p.G147E Chr.20: 46687075-46687075 on Build GRCh38
3294707 c.461A>G p.H154R Chr.20: 46687054-46687054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11550540 Chr.20: 46687130-46687130 on Build GRCh38
rs777533820 Chr.20: 46687036-46687036 on Build GRCh38
rs753763955 Chr.20: 46687176-46687176 on Build GRCh38
rs528622355 Chr.20: 46687127-46687127 on Build GRCh38
rs763202340 Chr.20: 46687135-46687135 on Build GRCh38
rs770369933 Chr.20: 46687053-46687053 on Build GRCh38
rs45570735 Chr.20: 46687178-46687178 on Build GRCh38
rs201387778 Chr.20: 46687215-46687215 on Build GRCh38
rs762402703 Chr.20: 46687221-46687221 on Build GRCh38
rs985927085 Chr.20: 46687043-46687043 on Build GRCh38
rs369690944 Chr.20: 46687216-46687216 on Build GRCh38
rs61729174 Chr.20: 46687111-46687111 on Build GRCh38
rs755798850 Chr.20: 46687160-46687160 on Build GRCh38
rs751828263 Chr.20: 46687151-46687151 on Build GRCh38
rs745434503 Chr.20: 46687186-46687186 on Build GRCh38
rs948899461 Chr.20: 46687181-46687181 on Build GRCh38
rs777191510 Chr.20: 46687218-46687218 on Build GRCh38
rs773296506 Chr.20: 46687198-46687199 on Build GRCh38
rs778452581 Chr.20: 46687074-46687074 on Build GRCh38
rs775905582 Chr.20: 46687086-46687086 on Build GRCh38
rs771666008 Chr.20: 46687083-46687083 on Build GRCh38
rs149404267 Chr.20: 46687060-46687060 on Build GRCh38
rs777637012 Chr.20: 46687171-46687171 on Build GRCh38
rs771638240 Chr.20: 46687189-46687189 on Build GRCh38
rs769092427 Chr.20: 46687096-46687096 on Build GRCh38
rs548440276 Chr.20: 46687196-46687196 on Build GRCh38
rs777207968 Chr.20: 46687109-46687109 on Build GRCh38
rs34983477 Chr.20: 46687147-46687147 on Build GRCh38
rs56008408 Chr.20: 46687082-46687082 on Build GRCh38
rs760903425 Chr.20: 46687092-46687092 on Build GRCh38
rs532864581 Chr.20: 46687177-46687177 on Build GRCh38
rs749230227 Chr.20: 46687052-46687052 on Build GRCh38
rs915072371 Chr.20: 46687194-46687194 on Build GRCh38
rs148722672 Chr.20: 46687198-46687198 on Build GRCh38
rs964818155 Chr.20: 46687157-46687157 on Build GRCh38

View Full Product Details