Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45924347-45924620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LARGE2
Entrez Gene ID
120071
Gene Name
LARGE xylosyl- and glucuronyltransferase 2
Gene Aliases
GYLTL1B, PP5656
UniGene
Hs.86543
Gene Chromosome Location
Chr.11: 45921621-45929096 on Build GRCh38
Transcripts
NM_001300721.1 NM_001300722.1 NM_152312.4 XM_005252787.2 XM_006718140.1 XM_006718141.3 XM_011519886.1 XM_011519888.1 XM_011519889.1 XM_011519890.1 XM_011519891.1 XM_011519892.2 XM_011519893.1 XM_017017173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
927096 c.586C>A p.R196S Chr.11: 45924599-45924599 on Build GRCh38
1746238 c.591C>T p.V197V Chr.11: 45924604-45924604 on Build GRCh38
2151555 c.543A>C p.L181L Chr.11: 45924556-45924556 on Build GRCh38
6677232 c.588C>T p.R196R Chr.11: 45924601-45924601 on Build GRCh38
289934 c.552G>C p.L184L Chr.11: 45924565-45924565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749785925 Chr.11: 45924486-45924486 on Build GRCh38
rs370021898 Chr.11: 45924547-45924547 on Build GRCh38
rs752491288 Chr.11: 45924544-45924544 on Build GRCh38
rs776510436 Chr.11: 45924516-45924516 on Build GRCh38
rs575347152 Chr.11: 45924541-45924541 on Build GRCh38
rs200282189 Chr.11: 45924520-45924520 on Build GRCh38
rs768356234 Chr.11: 45924601-45924601 on Build GRCh38
rs377634656 Chr.11: 45924542-45924542 on Build GRCh38
rs746840891 Chr.11: 45924511-45924511 on Build GRCh38
rs746673024 Chr.11: 45924600-45924600 on Build GRCh38
rs200854034 Chr.11: 45924545-45924545 on Build GRCh38
rs969623944 Chr.11: 45924489-45924489 on Build GRCh38
rs1029427913 Chr.11: 45924373-45924373 on Build GRCh38
rs757952553 Chr.11: 45924594-45924594 on Build GRCh38
rs931873733 Chr.11: 45924578-45924578 on Build GRCh38
rs767764254 Chr.11: 45924532-45924532 on Build GRCh38
rs751525688 Chr.11: 45924466-45924466 on Build GRCh38
rs376710862 Chr.11: 45924479-45924479 on Build GRCh38
rs767674499 Chr.11: 45924468-45924468 on Build GRCh38
rs949613073 Chr.11: 45924444-45924444 on Build GRCh38
rs144027171 Chr.11: 45924528-45924528 on Build GRCh38
rs553199792 Chr.11: 45924512-45924512 on Build GRCh38
rs1015045535 Chr.11: 45924411-45924411 on Build GRCh38
rs1050394623 Chr.11: 45924579-45924579 on Build GRCh38
rs1006705387 Chr.11: 45924403-45924403 on Build GRCh38
rs201899150 Chr.11: 45924599-45924599 on Build GRCh38
rs887808558 Chr.11: 45924390-45924390 on Build GRCh38
rs755257928 Chr.11: 45924467-45924467 on Build GRCh38
rs752907441 Chr.11: 45924538-45924538 on Build GRCh38
rs775261700 Chr.11: 45924506-45924506 on Build GRCh38
rs931507165 Chr.11: 45924496-45924496 on Build GRCh38
rs992291971 Chr.11: 45924447-45924447 on Build GRCh38
rs138459534 Chr.11: 45924523-45924523 on Build GRCh38
rs369644747 Chr.11: 45924499-45924499 on Build GRCh38
rs374032175 Chr.11: 45924529-45924529 on Build GRCh38
rs757674462 Chr.11: 45924465-45924465 on Build GRCh38
rs887768124 Chr.11: 45924593-45924593 on Build GRCh38
rs184765819 Chr.11: 45924406-45924406 on Build GRCh38
rs992867296 Chr.11: 45924399-45924399 on Build GRCh38
rs759324442 Chr.11: 45924461-45924462 on Build GRCh38
rs756435887 Chr.11: 45924472-45924472 on Build GRCh38
rs757650680 Chr.11: 45924490-45924490 on Build GRCh38
rs534314905 Chr.11: 45924477-45924477 on Build GRCh38
rs778492429 Chr.11: 45924495-45924495 on Build GRCh38
rs765214518 Chr.11: 45924502-45924527 on Build GRCh38
rs72902492 Chr.11: 45924418-45924418 on Build GRCh38

View Full Product Details