Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33242356-33242629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFA1
Entrez Gene ID
51297
Gene Name
BPI fold containing family A member 1
Gene Aliases
LUNX, NASG, PLUNC, SPLUNC1, SPURT, bA49G10.5
UniGene
Hs.211092
Gene Chromosome Location
Chr.20: 33235996-33243309 on Build GRCh38
Transcripts
NM_001243193.1 NM_016583.3 NM_130852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7169688 c.745G>C p.G249R Chr.20: 33242501-33242501 on Build GRCh38
6612324 c.762C>G p.I254M Chr.20: 33242518-33242518 on Build GRCh38
6317704 c.745G>T p.G249* Chr.20: 33242501-33242501 on Build GRCh38
6525738 c.744C>A p.H248Q Chr.20: 33242500-33242500 on Build GRCh38
7172872 c.753G>C p.Q251H Chr.20: 33242509-33242509 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1056362325 Chr.20: 33242434-33242434 on Build GRCh38
rs772839337 Chr.20: 33242490-33242490 on Build GRCh38
rs776757921 Chr.20: 33242403-33242404 on Build GRCh38
rs563479994 Chr.20: 33242593-33242593 on Build GRCh38
rs759939799 Chr.20: 33242500-33242500 on Build GRCh38
rs781338628 Chr.20: 33242456-33242456 on Build GRCh38
rs878875085 Chr.20: 33242498-33242498 on Build GRCh38
rs761697181 Chr.20: 33242437-33242437 on Build GRCh38
rs1044859958 Chr.20: 33242455-33242455 on Build GRCh38
rs776172316 Chr.20: 33242501-33242501 on Build GRCh38
rs757609021 Chr.20: 33242530-33242530 on Build GRCh38
rs377322301 Chr.20: 33242551-33242551 on Build GRCh38
rs929636330 Chr.20: 33242399-33242399 on Build GRCh38
rs983005111 Chr.20: 33242409-33242409 on Build GRCh38
rs892300828 Chr.20: 33242588-33242588 on Build GRCh38
rs1027049797 Chr.20: 33242383-33242383 on Build GRCh38
rs368602608 Chr.20: 33242559-33242560 on Build GRCh38
rs778104583 Chr.20: 33242453-33242453 on Build GRCh38
rs749039019 Chr.20: 33242473-33242473 on Build GRCh38
rs751983413 Chr.20: 33242445-33242446 on Build GRCh38
rs996557001 Chr.20: 33242377-33242377 on Build GRCh38
rs753156185 Chr.20: 33242442-33242442 on Build GRCh38
rs771476301 Chr.20: 33242486-33242486 on Build GRCh38
rs756252206 Chr.20: 33242578-33242578 on Build GRCh38
rs767880749 Chr.20: 33242466-33242467 on Build GRCh38
rs953738366 Chr.20: 33242604-33242604 on Build GRCh38
rs773934981 Chr.20: 33242477-33242477 on Build GRCh38
rs747802082 Chr.20: 33242485-33242485 on Build GRCh38
rs868767114 Chr.20: 33242561-33242561 on Build GRCh38
rs745777862 Chr.20: 33242466-33242466 on Build GRCh38
rs758663330 Chr.20: 33242445-33242445 on Build GRCh38
rs750988578 Chr.20: 33242469-33242470 on Build GRCh38
rs1052480535 Chr.20: 33242521-33242521 on Build GRCh38
rs953012890 Chr.20: 33242404-33242404 on Build GRCh38
rs749236339 Chr.20: 33242575-33242575 on Build GRCh38
rs146285061 Chr.20: 33242435-33242435 on Build GRCh38
rs185041484 Chr.20: 33242454-33242454 on Build GRCh38
rs750552385 Chr.20: 33242540-33242540 on Build GRCh38
rs372995591 Chr.20: 33242507-33242507 on Build GRCh38
rs768596583 Chr.20: 33242379-33242379 on Build GRCh38
rs764569180 Chr.20: 33242522-33242522 on Build GRCh38
rs918220166 Chr.20: 33242390-33242390 on Build GRCh38
rs377411186 Chr.20: 33242436-33242436 on Build GRCh38
rs1052268552 Chr.20: 33242541-33242541 on Build GRCh38
rs908718768 Chr.20: 33242410-33242410 on Build GRCh38
rs779992912 Chr.20: 33242557-33242557 on Build GRCh38
rs757661494 Chr.20: 33242466-33242470 on Build GRCh38
rs779678873 Chr.20: 33242470-33242470 on Build GRCh38
rs756123977 Chr.20: 33242469-33242469 on Build GRCh38
rs767469503 Chr.20: 33242535-33242535 on Build GRCh38
rs540978019 Chr.20: 33242389-33242389 on Build GRCh38

View Full Product Details