Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 45443257-45443496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX5
Entrez Gene ID
240
Gene Name
arachidonate 5-lipoxygenase
Gene Aliases
5-LO, 5-LOX, 5LPG, LOG5
UniGene
Hs.89499
Gene Chromosome Location
Chr.10: 45374166-45446121 on Build GRCh38
Transcripts
NM_000698.4 NM_001256153.2 NM_001256154.2 NM_001320861.1 NM_001320862.1 XM_017016012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6598710 c.1505A>G p.E502G Chr.10: 45443469-45443469 on Build GRCh38
5721490 c.1486G>A p.G496S Chr.10: 45443450-45443450 on Build GRCh38
4900148 c.1487G>A p.G496D Chr.10: 45443451-45443451 on Build GRCh38
4528363 c.1530G>A p.V510V Chr.10: 45443494-45443494 on Build GRCh38
1244860 c.1511C>T p.P504L Chr.10: 45443475-45443475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773636671 Chr.10: 45443393-45443393 on Build GRCh38
rs957592774 Chr.10: 45443346-45443346 on Build GRCh38
rs570263742 Chr.10: 45443340-45443340 on Build GRCh38
rs554497234 Chr.10: 45443441-45443441 on Build GRCh38
rs1051713 Chr.10: 45443298-45443298 on Build GRCh38
rs756915145 Chr.10: 45443449-45443449 on Build GRCh38
rs757919912 Chr.10: 45443457-45443457 on Build GRCh38
rs373344486 Chr.10: 45443469-45443469 on Build GRCh38
rs1031784148 Chr.10: 45443345-45443345 on Build GRCh38
rs746902367 Chr.10: 45443462-45443462 on Build GRCh38
rs761017436 Chr.10: 45443401-45443401 on Build GRCh38
rs904628194 Chr.10: 45443303-45443303 on Build GRCh38
rs199542576 Chr.10: 45443423-45443423 on Build GRCh38
rs776942877 Chr.10: 45443419-45443419 on Build GRCh38
rs986600875 Chr.10: 45443409-45443409 on Build GRCh38
rs537569092 Chr.10: 45443341-45443341 on Build GRCh38
rs895013345 Chr.10: 45443318-45443318 on Build GRCh38
rs772200618 Chr.10: 45443383-45443383 on Build GRCh38
rs372612423 Chr.10: 45443387-45443387 on Build GRCh38
rs762183261 Chr.10: 45443464-45443466 on Build GRCh38
rs769437415 Chr.10: 45443407-45443408 on Build GRCh38
rs1013498719 Chr.10: 45443337-45443337 on Build GRCh38
rs1013005022 Chr.10: 45443332-45443332 on Build GRCh38
rs766877023 Chr.10: 45443437-45443437 on Build GRCh38
rs965936120 Chr.10: 45443422-45443422 on Build GRCh38
rs921142800 Chr.10: 45443368-45443368 on Build GRCh38
rs55854006 Chr.10: 45443460-45443460 on Build GRCh38
rs370640705 Chr.10: 45443450-45443450 on Build GRCh38
rs749902144 Chr.10: 45443451-45443451 on Build GRCh38
rs770033748 Chr.10: 45443426-45443426 on Build GRCh38
rs913200890 Chr.10: 45443278-45443278 on Build GRCh38
rs746810158 Chr.10: 45443402-45443402 on Build GRCh38
rs772271784 Chr.10: 45443304-45443304 on Build GRCh38
rs143257667 Chr.10: 45443475-45443475 on Build GRCh38
rs745451578 Chr.10: 45443378-45443395 on Build GRCh38
rs755270606 Chr.10: 45443365-45443365 on Build GRCh38
rs753388561 Chr.10: 45443367-45443367 on Build GRCh38
rs773224245 Chr.10: 45443427-45443427 on Build GRCh38
rs549432246 Chr.10: 45443412-45443412 on Build GRCh38
rs774655444 Chr.10: 45443413-45443414 on Build GRCh38
rs777420174 Chr.10: 45443366-45443366 on Build GRCh38
rs375366789 Chr.10: 45443370-45443370 on Build GRCh38
rs913502418 Chr.10: 45443442-45443442 on Build GRCh38
rs1054950197 Chr.10: 45443317-45443317 on Build GRCh38
rs768682288 Chr.10: 45443467-45443467 on Build GRCh38
rs776670364 Chr.10: 45443344-45443344 on Build GRCh38
rs376874561 Chr.10: 45443425-45443425 on Build GRCh38
rs753078437 Chr.10: 45443443-45443443 on Build GRCh38
rs1002052221 Chr.10: 45443315-45443315 on Build GRCh38
rs763018509 Chr.10: 45443431-45443431 on Build GRCh38
rs199657122 Chr.10: 45443438-45443438 on Build GRCh38
rs745366345 Chr.10: 45443379-45443379 on Build GRCh38
rs993852240 Chr.10: 45443360-45443360 on Build GRCh38
rs572643270 Chr.10: 45443459-45443459 on Build GRCh38
rs894735671 Chr.10: 45443324-45443324 on Build GRCh38
rs748334405 Chr.10: 45443299-45443299 on Build GRCh38

View Full Product Details