Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 61868555-61868828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT4
Entrez Gene ID
10575
Gene Name
chaperonin containing TCP1 subunit 4
Gene Aliases
CCT-DELTA, Cctd, SRB
UniGene
Hs.421509
Gene Chromosome Location
Chr.2: 61868127-61888671 on Build GRCh38
Transcripts
NM_001256721.1 NM_006430.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7287873 c.1606-4T>A p.? Chr.2: 61868710-61868710 on Build GRCh38
4958317 c.1610A>C p.N537T Chr.2: 61868702-61868702 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1000824078 Chr.2: 61868742-61868742 on Build GRCh38
rs545854494 Chr.2: 61868631-61868631 on Build GRCh38
rs778819197 Chr.2: 61868676-61868677 on Build GRCh38
rs1023280948 Chr.2: 61868675-61868675 on Build GRCh38
rs938676952 Chr.2: 61868735-61868735 on Build GRCh38
rs757774567 Chr.2: 61868710-61868710 on Build GRCh38
rs750072250 Chr.2: 61868739-61868740 on Build GRCh38
rs373853411 Chr.2: 61868718-61868718 on Build GRCh38
rs756992819 Chr.2: 61868675-61868676 on Build GRCh38
rs749505745 Chr.2: 61868739-61868739 on Build GRCh38
rs764743443 Chr.2: 61868644-61868644 on Build GRCh38
rs745888201 Chr.2: 61868733-61868733 on Build GRCh38
rs199950866 Chr.2: 61868657-61868657 on Build GRCh38
rs764542502 Chr.2: 61868696-61868696 on Build GRCh38
rs772930866 Chr.2: 61868757-61868757 on Build GRCh38
rs991926329 Chr.2: 61868792-61868792 on Build GRCh38
rs760885270 Chr.2: 61868697-61868697 on Build GRCh38
rs758195762 Chr.2: 61868670-61868670 on Build GRCh38
rs1013276019 Chr.2: 61868665-61868665 on Build GRCh38
rs756195811 Chr.2: 61868738-61868738 on Build GRCh38
rs1032680868 Chr.2: 61868761-61868761 on Build GRCh38
rs896254332 Chr.2: 61868639-61868639 on Build GRCh38
rs758132724 Chr.2: 61868577-61868577 on Build GRCh38
rs768922366 Chr.2: 61868756-61868756 on Build GRCh38

View Full Product Details