Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 42759047-42759286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC152
Entrez Gene ID
100129792
Gene Name
coiled-coil domain containing 152
Gene Aliases
CH5400
UniGene
Hs.745017
Gene Chromosome Location
Chr.5: 42756818-42802437 on Build GRCh38
Transcripts
NM_001134848.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5953004 c.45G>C p.V15V Chr.5: 42759166-42759166 on Build GRCh38
1437648 c.56A>G p.K19R Chr.5: 42759177-42759177 on Build GRCh38
5587430 c.27G>A p.M9I Chr.5: 42759148-42759148 on Build GRCh38
1068360 c.72C>A p.F24L Chr.5: 42759193-42759193 on Build GRCh38
5522316 c.33G>A p.K11K Chr.5: 42759154-42759154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988119091 Chr.5: 42759146-42759146 on Build GRCh38
rs759445811 Chr.5: 42759177-42759177 on Build GRCh38
rs200364347 Chr.5: 42759193-42759193 on Build GRCh38
rs560359026 Chr.5: 42759257-42759257 on Build GRCh38
rs551978424 Chr.5: 42759237-42759237 on Build GRCh38
rs909776811 Chr.5: 42759242-42759242 on Build GRCh38
rs770647923 Chr.5: 42759163-42759163 on Build GRCh38
rs926438828 Chr.5: 42759234-42759234 on Build GRCh38
rs1008219424 Chr.5: 42759075-42759075 on Build GRCh38
rs751734058 Chr.5: 42759243-42759243 on Build GRCh38
rs183320572 Chr.5: 42759249-42759249 on Build GRCh38
rs1023213673 Chr.5: 42759162-42759162 on Build GRCh38
rs1036643020 Chr.5: 42759260-42759260 on Build GRCh38
rs781646954 Chr.5: 42759258-42759258 on Build GRCh38
rs1034705555 Chr.5: 42759122-42759122 on Build GRCh38
rs973568578 Chr.5: 42759201-42759201 on Build GRCh38
rs983917407 Chr.5: 42759240-42759240 on Build GRCh38
rs190380038 Chr.5: 42759239-42759239 on Build GRCh38
rs547819090 Chr.5: 42759083-42759083 on Build GRCh38
rs776583033 Chr.5: 42759173-42759173 on Build GRCh38
rs902264756 Chr.5: 42759085-42759085 on Build GRCh38
rs147430130 Chr.5: 42759157-42759157 on Build GRCh38
rs201689715 Chr.5: 42759128-42759128 on Build GRCh38
rs752714423 Chr.5: 42759209-42759209 on Build GRCh38
rs999651086 Chr.5: 42759121-42759121 on Build GRCh38
rs763769185 Chr.5: 42759247-42759247 on Build GRCh38

View Full Product Details