Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 41415281-41415506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD6
Entrez Gene ID
84181
Gene Name
chromodomain helicase DNA binding protein 6
Gene Aliases
CHD-6, CHD5, RIGB
UniGene
Hs.740645
Gene Chromosome Location
Chr.20: 41402101-41618494 on Build GRCh38
Transcripts
NM_032221.4 XM_005260573.2 XM_005260576.4 XM_011529080.2 XM_011529082.2 XM_017028099.1 XM_017028100.1 XM_017028101.1 XM_017028102.1 XM_017028103.1 XM_017028104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
13856 c.6679C>T p.P2227S Chr.20: 41415446-41415446 on Build GRCh38
3799546 c.6659C>G p.S2220C Chr.20: 41415466-41415466 on Build GRCh38
6628108 c.6638G>T p.G2213V Chr.20: 41415487-41415487 on Build GRCh38
1726402 c.6835A>G p.R2279G Chr.20: 41415290-41415290 on Build GRCh38
4504561 c.6693C>T p.S2231S Chr.20: 41415432-41415432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761666722 Chr.20: 41415455-41415455 on Build GRCh38
rs145690025 Chr.20: 41415346-41415346 on Build GRCh38
rs140533437 Chr.20: 41415342-41415342 on Build GRCh38
rs971071779 Chr.20: 41415343-41415343 on Build GRCh38
rs1040138327 Chr.20: 41415312-41415312 on Build GRCh38
rs771649762 Chr.20: 41415407-41415407 on Build GRCh38
rs202029350 Chr.20: 41415451-41415451 on Build GRCh38
rs756401605 Chr.20: 41415461-41415463 on Build GRCh38
rs147740968 Chr.20: 41415481-41415481 on Build GRCh38
rs534005191 Chr.20: 41415365-41415365 on Build GRCh38
rs1005786430 Chr.20: 41415422-41415422 on Build GRCh38
rs765014113 Chr.20: 41415466-41415466 on Build GRCh38
rs951814095 Chr.20: 41415417-41415417 on Build GRCh38
rs778008576 Chr.20: 41415395-41415395 on Build GRCh38
rs755155128 Chr.20: 41415349-41415349 on Build GRCh38
rs201404597 Chr.20: 41415415-41415415 on Build GRCh38
rs377662210 Chr.20: 41415450-41415450 on Build GRCh38
rs750214708 Chr.20: 41415476-41415476 on Build GRCh38
rs745496600 Chr.20: 41415400-41415400 on Build GRCh38
rs563580387 Chr.20: 41415325-41415325 on Build GRCh38
rs750599142 Chr.20: 41415308-41415308 on Build GRCh38
rs991222687 Chr.20: 41415337-41415337 on Build GRCh38
rs760147001 Chr.20: 41415416-41415416 on Build GRCh38
rs200490284 Chr.20: 41415333-41415333 on Build GRCh38
rs748667726 Chr.20: 41415389-41415389 on Build GRCh38
rs770140073 Chr.20: 41415392-41415392 on Build GRCh38

View Full Product Details