Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42249785-42250016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERF
Entrez Gene ID
2077
Gene Name
ETS2 repressor factor
Gene Aliases
CRS4, PE-2, PE2
UniGene
Hs.655969
Gene Chromosome Location
Chr.19: 42247561-42255164 on Build GRCh38
Transcripts
NM_001301035.1 NM_001308402.1 NM_001312656.1 NM_006494.3 XM_017026468.1 XM_017026469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5573661 c.258-2A>G p.? Chr.19: 42249944-42249944 on Build GRCh38
5190537 c.301C>T p.R101W Chr.19: 42249899-42249899 on Build GRCh38
997295 c.275G>A p.R92H Chr.19: 42249925-42249925 on Build GRCh38
6656061 c.302G>A p.R101Q Chr.19: 42249898-42249898 on Build GRCh38
6656060 c.310T>C p.Y104H Chr.19: 42249890-42249890 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs544806380 Chr.19: 42249990-42249990 on Build GRCh38
rs1045254229 Chr.19: 42249915-42249915 on Build GRCh38
rs774456067 Chr.19: 42249980-42249981 on Build GRCh38
rs556023808 Chr.19: 42249939-42249939 on Build GRCh38
rs769239795 Chr.19: 42249899-42249899 on Build GRCh38
rs747830281 Chr.19: 42249889-42249889 on Build GRCh38
rs746165064 Chr.19: 42249846-42249846 on Build GRCh38
rs565631783 Chr.19: 42249927-42249927 on Build GRCh38
rs764099591 Chr.19: 42249965-42249965 on Build GRCh38
rs886041001 Chr.19: 42249934-42249934 on Build GRCh38
rs904084126 Chr.19: 42249917-42249917 on Build GRCh38
rs140522983 Chr.19: 42249936-42249936 on Build GRCh38
rs762831348 Chr.19: 42249918-42249918 on Build GRCh38
rs1052030393 Chr.19: 42249937-42249937 on Build GRCh38
rs1033586611 Chr.19: 42249843-42249843 on Build GRCh38
rs760926921 Chr.19: 42249963-42249963 on Build GRCh38
rs79269780 Chr.19: 42249910-42249910 on Build GRCh38
rs201871955 Chr.19: 42249980-42249980 on Build GRCh38
rs780879505 Chr.19: 42249873-42249873 on Build GRCh38
rs753911446 Chr.19: 42249976-42249976 on Build GRCh38
rs756218487 Chr.19: 42249810-42249810 on Build GRCh38
rs770925496 Chr.19: 42249920-42249920 on Build GRCh38
rs368794183 Chr.19: 42249819-42249819 on Build GRCh38
rs772731129 Chr.19: 42249900-42249900 on Build GRCh38
rs750915227 Chr.19: 42249994-42249994 on Build GRCh38
rs774305612 Chr.19: 42249926-42249926 on Build GRCh38
rs768310421 Chr.19: 42249852-42249852 on Build GRCh38
rs866871043 Chr.19: 42249958-42249958 on Build GRCh38
rs754348493 Chr.19: 42249828-42249828 on Build GRCh38
rs983523826 Chr.19: 42249993-42249993 on Build GRCh38
rs757730703 Chr.19: 42249838-42249838 on Build GRCh38
rs779276597 Chr.19: 42249839-42249839 on Build GRCh38

View Full Product Details