Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78698749-78699022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH1
Entrez Gene ID
9267
Gene Name
cytohesin 1
Gene Aliases
B2-1, CYTOHESIN-1, D17S811E, PSCD1, SEC7
UniGene
Hs.191215
Gene Chromosome Location
Chr.17: 78674047-78782342 on Build GRCh38
Transcripts
NM_001292018.1 NM_001292019.1 NM_004762.3 NM_017456.3 XM_011525475.2 XM_011525476.2 XM_011525477.1 XM_011525478.1 XM_017025336.1 XM_017025337.1 XM_017025338.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4502229 c.608C>A p.P203H Chr.17: 78698911-78698911 on Build GRCh38
6233391 c.683C>A p.P228Q Chr.17: 78698836-78698836 on Build GRCh38
6148517 c.600G>T p.L200L Chr.17: 78698919-78698919 on Build GRCh38
985176 c.642C>A p.F214L Chr.17: 78698877-78698877 on Build GRCh38
6639536 c.561C>T p.Y187Y Chr.17: 78698958-78698958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930764625 Chr.17: 78698789-78698789 on Build GRCh38
rs772507196 Chr.17: 78698873-78698873 on Build GRCh38
rs774719566 Chr.17: 78698787-78698787 on Build GRCh38
rs754087658 Chr.17: 78698821-78698821 on Build GRCh38
rs757417957 Chr.17: 78698822-78698822 on Build GRCh38
rs765492162 Chr.17: 78698957-78698957 on Build GRCh38
rs997505651 Chr.17: 78698853-78698853 on Build GRCh38
rs150354793 Chr.17: 78698958-78698958 on Build GRCh38
rs776630136 Chr.17: 78698934-78698934 on Build GRCh38
rs201122386 Chr.17: 78698835-78698835 on Build GRCh38
rs766238099 Chr.17: 78698986-78698986 on Build GRCh38
rs574814614 Chr.17: 78698908-78698908 on Build GRCh38
rs762785415 Chr.17: 78698966-78698966 on Build GRCh38
rs751571554 Chr.17: 78698990-78698990 on Build GRCh38
rs141177293 Chr.17: 78698942-78698942 on Build GRCh38
rs758333238 Chr.17: 78698833-78698833 on Build GRCh38
rs900423792 Chr.17: 78698902-78698902 on Build GRCh38
rs372876719 Chr.17: 78698999-78698999 on Build GRCh38
rs372618002 Chr.17: 78698800-78698800 on Build GRCh38
rs780853745 Chr.17: 78698998-78698998 on Build GRCh38
rs768742179 Chr.17: 78698913-78698913 on Build GRCh38
rs138731073 Chr.17: 78698826-78698826 on Build GRCh38
rs746234210 Chr.17: 78698867-78698867 on Build GRCh38
rs775941917 Chr.17: 78698904-78698904 on Build GRCh38
rs369357942 Chr.17: 78698854-78698854 on Build GRCh38
rs750164978 Chr.17: 78698829-78698829 on Build GRCh38
rs760719589 Chr.17: 78698801-78698801 on Build GRCh38
rs746427818 Chr.17: 78698850-78698850 on Build GRCh38
rs779215895 Chr.17: 78698863-78698863 on Build GRCh38
rs764337441 Chr.17: 78698811-78698811 on Build GRCh38
rs375897965 Chr.17: 78698836-78698836 on Build GRCh38
rs200100646 Chr.17: 78698991-78698991 on Build GRCh38
rs747817213 Chr.17: 78698858-78698858 on Build GRCh38
rs759970244 Chr.17: 78698790-78698790 on Build GRCh38
rs771222773 Chr.17: 78698774-78698774 on Build GRCh38
rs754567835 Chr.17: 78698842-78698842 on Build GRCh38

View Full Product Details