Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 122302730-122302999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEZF1
Entrez Gene ID
389549
Gene Name
FEZ family zinc finger 1
Gene Aliases
FEZ, HH22, ZNF312B
UniGene
Hs.553970
Gene Chromosome Location
Chr.7: 122300939-122311855 on Build GRCh38
Transcripts
NM_001024613.3 NM_001160264.2 XM_005250337.3 XM_011516202.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
744829 c.1011A>T p.I337I Chr.7: 122302857-122302857 on Build GRCh38
335251 c.1004C>T p.T335I Chr.7: 122302864-122302864 on Build GRCh38
6261343 c.986C>A p.S329Y Chr.7: 122302882-122302882 on Build GRCh38
5587821 c.987C>T p.S329S Chr.7: 122302881-122302881 on Build GRCh38
3684990 c.1002T>C p.H334H Chr.7: 122302866-122302866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs869285925 Chr.7: 122302782-122302783 on Build GRCh38
rs748444890 Chr.7: 122302972-122302973 on Build GRCh38
rs995750757 Chr.7: 122302946-122302946 on Build GRCh38
rs201126466 Chr.7: 122302777-122302777 on Build GRCh38
rs6968786 Chr.7: 122302793-122302793 on Build GRCh38
rs184114123 Chr.7: 122302760-122302760 on Build GRCh38
rs371893844 Chr.7: 122302968-122302968 on Build GRCh38
rs117528546 Chr.7: 122302795-122302795 on Build GRCh38
rs766475299 Chr.7: 122302851-122302851 on Build GRCh38
rs10685687 Chr.7: 122302778-122302779 on Build GRCh38
rs769231658 Chr.7: 122302925-122302925 on Build GRCh38
rs779053523 Chr.7: 122302870-122302870 on Build GRCh38
rs200646623 Chr.7: 122302879-122302879 on Build GRCh38
rs148805926 Chr.7: 122302848-122302848 on Build GRCh38
rs72569111 Chr.7: 122302780-122302781 on Build GRCh38
rs1036729192 Chr.7: 122302880-122302880 on Build GRCh38
rs368199722 Chr.7: 122302784-122302784 on Build GRCh38
rs758693318 Chr.7: 122302902-122302902 on Build GRCh38
rs774082152 Chr.7: 122302952-122302952 on Build GRCh38
rs775046683 Chr.7: 122302926-122302926 on Build GRCh38
rs754046193 Chr.7: 122302852-122302852 on Build GRCh38
rs769335729 Chr.7: 122302790-122302790 on Build GRCh38
rs397826372 Chr.7: 122302781-122302782 on Build GRCh38
rs750318384 Chr.7: 122302840-122302840 on Build GRCh38
rs35985998 Chr.7: 122302779-122302780 on Build GRCh38
rs779217330 Chr.7: 122302763-122302763 on Build GRCh38
rs770466266 Chr.7: 122302766-122302770 on Build GRCh38
rs748758252 Chr.7: 122302939-122302939 on Build GRCh38
rs78309987 Chr.7: 122302866-122302866 on Build GRCh38
rs761716274 Chr.7: 122302955-122302955 on Build GRCh38
rs773725968 Chr.7: 122302785-122302785 on Build GRCh38
rs764033610 Chr.7: 122302818-122302818 on Build GRCh38
rs369157481 Chr.7: 122302941-122302941 on Build GRCh38
rs774309900 Chr.7: 122302820-122302820 on Build GRCh38
rs778154305 Chr.7: 122302918-122302918 on Build GRCh38
rs761661056 Chr.7: 122302824-122302824 on Build GRCh38
rs748698017 Chr.7: 122302774-122302774 on Build GRCh38
rs745371779 Chr.7: 122302787-122302787 on Build GRCh38

View Full Product Details