Species:
Dasypus novemcinctus, Guinea pig, Papio hamadryas, Pig, Rabbit
Mature miRNA Sequence:
CCCCCAGGUGUGAUUCUGAUUUGC
Mapped to miRBase Version:
v22.1

Stem-loop Details

Stem-loop ID Stem-loop Accession # Mature miRNA ID Stem-loop Sequence Chromosome Location
cpo-mir-361 MI0038742 cpo-miR-361-3p UUAUCAGAAUCUCCAGGGGUACUUAUAAUUUGUAAAAGUCCCCCAGGUGUGAUUCUGAUUUGC -
dno-mir-361 MI0039079 dno-miR-361-3p UUAUCAGAAUCUCCAGGGGUACUUAAUAUGUGAAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGC -
ocu-mir-361 MI0039377 ocu-miR-361-3p UUAUCAGAAUCUCCAGGGGUACUUACAAUUUGAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGC -
pha-mir-361 MI0040276 pha-miR-361 UUAUCAGAAUCUCCAGGGGUACUUUAUAAUUUCAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGC -
ssc-mir-361 MI0013145 ssc-miR-361-3p GGAGCUUAUCAGAAUCUCCAGGGGUACUUAUAAUUUGAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGCUUCCUCCUCCUC -

View Full Stem-loop Details