Gene Transcripts

Gene Symbol
Slc14a1
Entrez Gene ID
108052
Gene Name
solute carrier family 14 (urea transporter), member 1
Gene Aliases
2610507K20Rik, 3021401A05Rik, UT-B, Utb1
Chromosome Location
Chr. 18: 78100091 - 78142119 on Build GRCm38
UniGene ID
Mm.33832
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171010.1 NP_001164481.1 4 568
NM_001171011.1 NP_001164482.1 3 303
NM_028122.4 NP_082398.1 3 363
GenBank mRNA AF448798.1 - 2 160
AJ420967.1 - 3 222
AK012066.1 - 3 213
AK041979.1 - 3 305
AK153891.1 - 4 473
BC058594.1 - 3 250 5364683
BC086673.1 - 3 259 30646772

View Full Product Details