Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001003911.2 | NP_001003911.2 | 6 | 1185 | |
GenBank mRNA | AY551090.1 | - | 6 | 918 | |
BC030442.1 | - | 1 | 46 | ||
BC058991.1 | - | 3 | 163 | ||
BC076575.1 | - | 6 | 1039 | ||
BC141173.1 | - | 6 | 961 | 9055785 | |
BC145491.1 | - | 6 | 961 | 9054054 |