Gene Transcripts

Gene Symbol
Glra3
Entrez Gene ID
110304
Gene Name
glycine receptor, alpha 3 subunit
Gene Aliases
Chromosome Location
Chr. 8: 55940411 - 56130260 on Build GRCm38
UniGene ID
Mm.307061
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080438.2 NP_536686.2 7 814
GenBank mRNA AF214575.1 - 3 216
AF362764.1 - 7 732
AY230204.1 - 7 753

View Full Product Details