Gene Transcripts

Gene Symbol
Egln2
Entrez Gene ID
112406
Gene Name
egl-9 family hypoxia-inducible factor 2
Gene Aliases
0610011A13Rik, C85656, Hif-p4h-1, Ier4, Phd1, SM-20
Chromosome Location
Chr. 7: 27158658 - 27166802 on Build GRCm38
UniGene ID
Mm.29978
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053208.4 NP_444438.2 3 1270
GenBank mRNA AF453879.1 - 3 1192
AJ310547.1 - 3 1241
AK075582.1 - 3 1222
AK152056.1 - 3 1231
AK152223.1 - 3 1228
AK159206.1 - 3 1223
AK166661.1 - 4 1136
AK167477.1 - 3 1223
BC023299.1 - 3 1204 5135579
BC051436.1 - 2 988 6312638
BC086764.1 - 3 1209 5685311

View Full Product Details