Gene Transcripts

Gene Symbol
Abca4
Entrez Gene ID
11304
Gene Name
ATP-binding cassette, sub-family A (ABC1), member 4
Gene Aliases
AW050280, Abc10, Abcr, D430003I15Rik, RmP
Chromosome Location
Chr. 3: 122043893 - 122180070 on Build GRCm38
UniGene ID
Mm.3918
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007378.1 NP_031404.1 27, 28 4204
GenBank mRNA AF000149.1 - 27, 28 4204
AK052916.1 - 4, 5 562
BC057853.1 - 27, 28 4204 6492139

View Full Product Details