Gene Transcripts

Gene Symbol
Adam5
Entrez Gene ID
11499
Gene Name
a disintegrin and metallopeptidase domain 5
Gene Aliases
tMDC II, tMDCII
Chromosome Location
Chr. 8: 24727093 - 24824369 on Build GRCm38
UniGene ID
Mm.143889
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001272057.1 NP_001258986.1 7 933
NM_001272058.1 NP_001258987.1 7 933
NM_001272059.1 NP_001258988.1 7 933
NM_007401.3 NP_031427.2 7 933
GenBank mRNA AK016574.1 - 7 589
AK029519.1 - 7 623
AK161262.1 - 7 933
AK161415.1 - 4 311
AK161417.1 - 7 603
BC094237.1 - 7 630 30604517
U22059.1 - 7 597

View Full Product Details