Gene Transcripts

Gene Symbol
Cops2
Entrez Gene ID
12848
Gene Name
COP9 (constitutive photomorphogenic) homolog, subunit 2 (Arabidopsis thaliana)
Gene Aliases
AI315723, C85265, Csn2, Sgn2, TRIP-15, Trip15
Chromosome Location
Chr. 2: 125830302 - 125859082 on Build GRCm38
UniGene ID
Mm.3596
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285507.1 NP_001272436.1 13 1431
NM_001285512.1 NP_001272441.1 13 1458
NM_001285513.1 NP_001272442.1 12 1222
NM_009939.3 NP_034069.2 13 1410
GenBank mRNA AF071312.1 - 13 1320
AF087688.1 - 13 1317
AK010383.1 - 13 1357
AK037757.1 - 13 1357
AK084421.1 - 13 1331
AK132207.1 - 13 1327
AK132776.1 - 13 1395
AK145225.1 - 12 1134
AK146153.1 - 13 1321
BC023096.1 - 13 1315 3675508

View Full Product Details