Gene Transcripts

Gene Symbol
Dnah8
Entrez Gene ID
13417
Gene Name
dynein, axonemal, heavy chain 8
Gene Aliases
ATPase, Dnahc8, Hst6.7b, P1-Loop
Chromosome Location
Chr. 17: 30626936 - 30875762 on Build GRCm38
UniGene ID
Mm.426017
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013811.3 NP_038839.2 13876
GenBank mRNA AF356520.1 - 13876
AF356522.1 - 13886
AK133238.1 - 19 2631

View Full Product Details