Gene Transcripts
- Gene Symbol
- Eif3a
- Entrez Gene ID
- 13669
- Gene Name
- eukaryotic translation initiation factor 3, subunit A
- Gene Aliases
- A830012B05Rik, Csma, Eif3, Eif3s10, mKIAA0139
- Chromosome Location
- Chr. 19: 60761116 - 60790693 on Build GRCm38
- UniGene ID
- Mm.2238
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010123.3 |
NP_034253.3
|
9, 10 |
1500 |
|
GenBank mRNA |
AK082006.1 |
- |
9, 10 |
1466 |
|
AK145132.1 |
- |
9, 10 |
1508 |
|
AK145629.1 |
- |
9, 10 |
1465 |
|
AK149482.1 |
- |
9, 10 |
1453 |
|
AK161634.1 |
- |
9, 10 |
1457 |
|
AK167010.1 |
- |
9, 10 |
1467 |
|
AK172899.1 |
- |
9, 10 |
1437 |
|
U14172.1 |
- |
9, 10 |
1450 |
|
X17373.1 |
- |
2, 3 |
124 |
|
X84651.1 |
- |
2, 3 |
124 |
|
View Full Product Details