Gene Transcripts

Gene Symbol
Epm2a
Entrez Gene ID
13853
Gene Name
epilepsy, progressive myoclonic epilepsy, type 2 gene alpha
Gene Aliases
TG-B, Tg(TcraK,TcrbK)TG-BFlv
Chromosome Location
Chr. 10: 11342690 - 11457477 on Build GRCm38
UniGene ID
Mm.89946
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010146.2 NP_034276.2 4 893
GenBank mRNA AF124044.1 - 4 933
CN532498.1 - 3 524

View Full Product Details