Gene Transcripts

Gene Symbol
F2rl2
Entrez Gene ID
14064
Gene Name
coagulation factor II (thrombin) receptor-like 2
Gene Aliases
F730031A08, PAR3
Chromosome Location
Chr. 13: 95696920 - 95702768 on Build GRCm38
UniGene ID
Mm.440952
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010170.4 NP_034300.3 2 996
GenBank mRNA AK089444.1 - 2 960
AK135212.1 - 2 1063
BC140985.1 - 2 951 9055597
U92972.1 - 2 1004

View Full Product Details