Gene Transcripts

Gene Symbol
Fzd3
Entrez Gene ID
14365
Gene Name
frizzled homolog 3 (Drosophila)
Gene Aliases
AU020229, D930050A07Rik, Fz3
Chromosome Location
Chr. 14: 65192441 - 65262463 on Build GRCm38
UniGene ID
Mm.214687
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021458.2 NP_067433.1 7 2338
GenBank mRNA BC050965.1 - 7 2299
BC066186.1 - 7 1998
BC103714.1 - 7 1998
U43205.1 - 6 2187

View Full Product Details