Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001109657.3 | NP_001103127.1 | 4 | 571 | |
NM_001277079.1 | NP_001264008.1 | 4 | 400 | ||
NM_008088.5 | NP_032114.2 | 4 | 377 | ||
GenBank mRNA | AK017394.1 | - | 4 | 371 | |
AK076926.1 | - | 5 | 453 | ||
AK152149.1 | - | 2 | 74 | ||
AK154463.1 | - | 4 | 374 | ||
BC120528.1 | - | 4 | 298 | 40129451 | |
BC120530.1 | - | 4 | 298 | 40129453 | |
BU053145.1 | - | 4 | 381 | ||
CJ147966.1 | - | 4 | 401 | ||
U19860.1 | - | 4 | 403 |