Gene Transcripts

Gene Symbol
Grin2d
Entrez Gene ID
14814
Gene Name
glutamate receptor, ionotropic, NMDA2D (epsilon 4)
Gene Aliases
GluN2D, NMDAR2D, NR2D
Chromosome Location
Chr. 7: 45831883 - 45872689 on Build GRCm38
UniGene ID
Mm.322594
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008172.2 NP_032198.2 8 2111
GenBank mRNA AB154244.1 - 8 1997
AB154245.1 - 8 1997
D12822.1 - 8 1997

View Full Product Details