Gene Transcripts

Gene Symbol
Gyk
Entrez Gene ID
14933
Gene Name
glycerol kinase
Gene Aliases
D930012N15Rik, GK
Chromosome Location
Chr. X: 85701937 - 85776819 on Build GRCm38
UniGene ID
Mm.246682
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001294138.1 NP_001281067.1 14 1402
NM_001294140.1 NP_001281069.1 14 1336
NM_008194.3 NP_032220.1 13 1318
NM_212444.2 NP_997609.1 13 1318
GenBank mRNA AK044308.1 - 14 1174
AK088373.1 - 13 1137
AK169424.1 - 14 1208
AK169525.1 - 13 1319
BC003767.1 - 13 1121 3593896
BQ555961.1 - 3 183
CB950214.1 - 8 500
CK789590.1 - 8 468
U48403.1 - 13 1095

View Full Product Details