Gene Transcripts

Gene Symbol
Kif11
Entrez Gene ID
16551
Gene Name
kinesin family member 11
Gene Aliases
Eg5, Kif8, Kifl1, Knsl1
Chromosome Location
Chr. 19: 37376403 - 37421859 on Build GRCm38
UniGene ID
Mm.42203
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010615.1 NP_034745.1 3 444
GenBank mRNA AJ223293.1 - 2 142
AK083324.1 - 3 420
AK147754.1 - 3 467
BC060670.1 - 3 444 6406754

View Full Product Details