Gene Transcripts

Gene Symbol
Lrrc23
Entrez Gene ID
16977
Gene Name
leucine rich repeat containing 23
Gene Aliases
4921537K05Rik, B7, Lrpb7
Chromosome Location
Chr. 6: 124769869 - 124780341 on Build GRCm38
UniGene ID
Mm.3207
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK015011.1 - 6, 7 836
BC049601.1 - 6, 7 844 6743951

View Full Product Details