Gene Transcripts

Gene Symbol
Trpm8
Entrez Gene ID
171382
Gene Name
transient receptor potential cation channel, subfamily M, member 8
Gene Aliases
CMR1, LTRPC6, LTrpC-6, TRPP8, Trp-p8
Chromosome Location
Chr. 1: 88306712 - 88388853 on Build GRCm38
UniGene ID
Mm.218753
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134252.3 NP_599013.1 6 673
GenBank mRNA AF481480.2 - 942
AY095352.1 - 4 507
BC117934.1 - 6 673 40097778

View Full Product Details