Gene Transcripts

Gene Symbol
Mak
Entrez Gene ID
17152
Gene Name
male germ cell-associated kinase
Gene Aliases
A930010O05Rik
Chromosome Location
Chr. 13: 41025007 - 41079706 on Build GRCm38
UniGene ID
Mm.8149
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145803.1 NP_001139275.1 14 2038
NM_008547.2 NP_032573.2 12 1861
GenBank mRNA X66983.1 - 12 1855

View Full Product Details