Gene Transcripts
- Gene Symbol
- Mcm5
- Entrez Gene ID
- 17218
- Gene Name
- minichromosome maintenance deficient 5, cell division cycle 46 (S. cerevisiae)
- Gene Aliases
- AA617332, AI324988, AL033333, Cdc46, Mcmd5, P1-CDC46
- Chromosome Location
- Chr. 8: 75109509 - 75128439 on Build GRCm38
- UniGene ID
- Mm.5048
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK033196.1 |
- |
9, 10 |
1273 |
|
AK051795.1 |
- |
9, 10 |
1243 |
|
AK088558.1 |
- |
2, 3 |
189 |
|
AK133826.1 |
- |
9, 10 |
1274 |
|
AK209639.1 |
- |
1, 2 |
129 |
|
AK212108.1 |
- |
1, 2 |
129 |
|
AK214383.1 |
- |
1, 2 |
129 |
|
BC090645.1 |
- |
9, 10 |
1226 |
|
BC094416.1 |
- |
9, 10 |
1238 |
5707724 |
D26090.1 |
- |
8, 9 |
1293 |
|
View Full Product Details