Gene Transcripts

Gene Symbol
Mgrn1
Entrez Gene ID
17237
Gene Name
mahogunin, ring finger 1
Gene Aliases
2610042J20Rik, mKIAA0544, md, nc
Chromosome Location
Chr. 16: 4886100 - 4938296 on Build GRCm38
UniGene ID
Mm.291326
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252437.1 NP_001239366.1 12 1402
NM_029657.4 NP_083933.1 12 1399
GenBank mRNA AK011747.2 - 12 1192
AK034100.1 - 3 230
AK088533.1 - 12 1182
AK129161.1 - 9 810
AK150176.1 - 13 1235
AK153407.1 - 12 1223
AK178579.1 - 3 192
AK179898.1 - 3 193
AK179943.1 - 248
AK182750.1 - 2 127
AK186073.1 - 2 126
AK187994.1 - 3 195
AK192679.1 - 2 126
AK193360.1 - 3 192
AK196471.1 - 3 192
AK200602.1 - 3 192
AK208234.1 - 2 126
AK209178.1 - 3 192
AK211037.1 - 2 126
AK211419.1 - 3 192
AK214313.1 - 3 192
AK214363.1 - 3 192
BC046830.1 - 12 1248 6811606

View Full Product Details