Gene Transcripts
- Gene Symbol
- Mmp14
- Entrez Gene ID
- 17387
- Gene Name
- matrix metallopeptidase 14 (membrane-inserted)
- Gene Aliases
- AI325305, MT-MMP-1, MT1-MMP, sabe
- Chromosome Location
- Chr. 14: 54431604 - 54441258 on Build GRCm38
- UniGene ID
- Mm.280175
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008608.3 |
NP_032634.3
|
9, 10 |
1677 |
|
GenBank mRNA |
AK088476.1 |
- |
9, 10 |
1677 |
|
AK134340.1 |
- |
9, 10 |
1609 |
|
AK138611.1 |
- |
9, 10 |
1618 |
|
AK149636.1 |
- |
9, 10 |
1606 |
|
AK149907.1 |
- |
9, 10 |
1657 |
|
AK149988.1 |
- |
9, 10 |
1577 |
|
AK155117.1 |
- |
9, 10 |
1679 |
|
AK163601.1 |
- |
9, 10 |
1669 |
|
AK165014.1 |
- |
9, 10 |
1678 |
|
AK185366.1 |
- |
2, 3 |
226 |
|
AK185474.1 |
- |
1, 2 |
107 |
|
AK186174.1 |
- |
1, 2 |
107 |
|
AK186619.1 |
- |
1, 2 |
107 |
|
AK189497.1 |
- |
2, 3 |
225 |
|
AK194864.1 |
- |
1, 2 |
107 |
|
AK212820.1 |
- |
1, 2 |
107 |
|
BC076638.1 |
- |
9, 10 |
1669 |
30658833 |
DQ249870.1 |
- |
9, 10 |
1415 |
|
U54984.1 |
- |
9, 10 |
1661 |
|
X83536.2 |
- |
9, 10 |
1587 |
|
View Full Product Details