Gene Transcripts
- Gene Symbol
- Nfkb1
- Entrez Gene ID
- 18033
- Gene Name
- nuclear factor of kappa light polypeptide gene enhancer in B cells 1, p105
- Gene Aliases
- NF-KB1, NF-kappaB, NF-kappaB1, p105, p50, p50/p105
- Chromosome Location
- Chr. 3: 135584655 - 135691547 on Build GRCm38
- UniGene ID
- Mm.256765
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008689.2 |
NP_032715.2
|
15, 16 |
1971 |
|
GenBank mRNA |
AB119195.1 |
- |
13, 14 |
1469 |
|
AB221614.1 |
- |
13, 14 |
1476 |
|
AK089660.1 |
- |
3, 4 |
345 |
|
AK146389.1 |
- |
1, 2 |
191 |
|
AK159423.1 |
- |
3, 4 |
402 |
|
AY521462.1 |
- |
13, 14 |
1469 |
|
AY521463.1 |
- |
13, 14 |
1469 |
|
BC138535.1 |
- |
14, 15 |
1548 |
8861555 |
BC138536.1 |
- |
14, 15 |
1548 |
8861556 |
BC145317.1 |
- |
15, 16 |
1565 |
|
L28117.1 |
- |
3, 4 |
385 |
|
M57999.1 |
- |
15, 16 |
1760 |
|
S66656.1 |
- |
3, 4 |
515 |
|
S89033.1 |
- |
3, 4 |
563 |
|
View Full Product Details