Gene Transcripts

Gene Symbol
Pabpc1
Entrez Gene ID
18458
Gene Name
poly(A) binding protein, cytoplasmic 1
Gene Aliases
PABP, Pabp1, PabpI, Pabpl1, ePAB
Chromosome Location
Chr. 15: 36595658 - 36609581 on Build GRCm38
UniGene ID
Mm.371570
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008774.3 NP_032800.2 10 1843
GenBank mRNA AK044829.1 - 10 1840
AK089747.1 - 10 1843
AK133196.1 - 10 1844
AK150839.1 - 10 1844
AK152064.1 - 10 1811
AK159703.1 - 10 1843
AK159733.1 - 10 1843
AK160883.1 - 10 1843
AK160968.1 - 10 1842
AK161123.1 - 10 1846
AK161671.1 - 10 1842
AK168466.1 - 10 1842
BC003870.1 - 10 1462 3491915
BC004587.1 - 9 1049
BC011207.1 - 10 1600 4022169
BC023145.1 - 10 1438 3988101
BC046233.1 - 10 1833 6816124
X65553.1 - 10 1629

View Full Product Details